Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24178431-24178690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
UniGene
Hs.61345
Gene Chromosome Location
Chr.6: 24171755-24383292 on Build GRCh38
Transcripts
NM_001195610.1 NM_016356.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326489 c.454G>A p.A152T Chr.6: 24178461-24178461 on Build GRCh38
1723937 c.1170G>A p.E390E Chr.6: 24178486-24178486 on Build GRCh38
4460419 c.423C>T p.V141V Chr.6: 24178492-24178492 on Build GRCh38
6482246 c.289G>T p.A97S Chr.6: 24178626-24178626 on Build GRCh38
6483189 c.443G>T p.S148I Chr.6: 24178472-24178472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765153120 Chr.6: 24178635-24178635 on Build GRCh38
rs150760955 Chr.6: 24178630-24178630 on Build GRCh38
rs75899525 Chr.6: 24178659-24178659 on Build GRCh38
rs773301531 Chr.6: 24178558-24178558 on Build GRCh38
rs1052135359 Chr.6: 24178582-24178582 on Build GRCh38
rs768048150 Chr.6: 24178486-24178486 on Build GRCh38
rs767129413 Chr.6: 24178492-24178492 on Build GRCh38
rs201827080 Chr.6: 24178547-24178547 on Build GRCh38
rs776956291 Chr.6: 24178527-24178532 on Build GRCh38
rs758852015 Chr.6: 24178452-24178452 on Build GRCh38
rs778095991 Chr.6: 24178548-24178548 on Build GRCh38
rs752761872 Chr.6: 24178524-24178524 on Build GRCh38
rs973946416 Chr.6: 24178485-24178485 on Build GRCh38
rs763068019 Chr.6: 24178654-24178654 on Build GRCh38
rs753636454 Chr.6: 24178632-24178632 on Build GRCh38
rs774111540 Chr.6: 24178490-24178490 on Build GRCh38
rs770279005 Chr.6: 24178586-24178586 on Build GRCh38
rs760502604 Chr.6: 24178614-24178614 on Build GRCh38
rs180988889 Chr.6: 24178509-24178509 on Build GRCh38
rs755149513 Chr.6: 24178521-24178521 on Build GRCh38
rs953735295 Chr.6: 24178505-24178505 on Build GRCh38
rs766600237 Chr.6: 24178628-24178628 on Build GRCh38
rs777259245 Chr.6: 24178458-24178458 on Build GRCh38
rs753739433 Chr.6: 24178514-24178514 on Build GRCh38
rs765215691 Chr.6: 24178522-24178522 on Build GRCh38
rs752022886 Chr.6: 24178554-24178554 on Build GRCh38
rs747339767 Chr.6: 24178454-24178454 on Build GRCh38
rs143313706 Chr.6: 24178612-24178612 on Build GRCh38
rs771591530 Chr.6: 24178457-24178457 on Build GRCh38
rs773020868 Chr.6: 24178502-24178502 on Build GRCh38
rs748310193 Chr.6: 24178451-24178451 on Build GRCh38
rs748873318 Chr.6: 24178474-24178474 on Build GRCh38
rs911570078 Chr.6: 24178546-24178546 on Build GRCh38
rs761388263 Chr.6: 24178491-24178491 on Build GRCh38
rs538198742 Chr.6: 24178557-24178557 on Build GRCh38
rs760412461 Chr.6: 24178503-24178503 on Build GRCh38
rs771461514 Chr.6: 24178512-24178513 on Build GRCh38
rs1002544728 Chr.6: 24178641-24178641 on Build GRCh38
rs143452599 Chr.6: 24178453-24178453 on Build GRCh38
rs905003012 Chr.6: 24178637-24178637 on Build GRCh38
rs747652700 Chr.6: 24178600-24178600 on Build GRCh38
rs762570866 Chr.6: 24178567-24178567 on Build GRCh38
rs1056349487 Chr.6: 24178647-24178647 on Build GRCh38
rs986382887 Chr.6: 24178520-24178520 on Build GRCh38
rs183480366 Chr.6: 24178590-24178590 on Build GRCh38
rs946583171 Chr.6: 24178611-24178611 on Build GRCh38
rs746347880 Chr.6: 24178585-24178585 on Build GRCh38
rs754765133 Chr.6: 24178648-24178648 on Build GRCh38
rs768017577 Chr.6: 24178654-24178654 on Build GRCh38
rs962387857 Chr.6: 24178501-24178501 on Build GRCh38
rs140580401 Chr.6: 24178555-24178555 on Build GRCh38
rs34647318 Chr.6: 24178475-24178475 on Build GRCh38
rs757670255 Chr.6: 24178556-24178556 on Build GRCh38
rs535914952 Chr.6: 24178619-24178619 on Build GRCh38
rs772827889 Chr.6: 24178613-24178613 on Build GRCh38

View Full Product Details