Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102602880-102603153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5
Entrez Gene ID
64860
Gene Name
armadillo repeat containing, X-linked 5
Gene Aliases
GASP5
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599168-102604159 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

SNP Detail

SNP ID Chromosome Location
rs765121989 Chr.X: 102603037-102603037 on Build GRCh38
rs760401300 Chr.X: 102603014-102603014 on Build GRCh38
rs887148982 Chr.X: 102603024-102603024 on Build GRCh38
rs113918916 Chr.X: 102603073-102603073 on Build GRCh38
rs937799597 Chr.X: 102602921-102602921 on Build GRCh38
rs759494842 Chr.X: 102603048-102603048 on Build GRCh38
rs747215363 Chr.X: 102602931-102602931 on Build GRCh38
rs752461022 Chr.X: 102602916-102602916 on Build GRCh38
rs781215205 Chr.X: 102602948-102602948 on Build GRCh38
rs759070944 Chr.X: 102603026-102603026 on Build GRCh38
rs772797845 Chr.X: 102603012-102603012 on Build GRCh38
rs376270648 Chr.X: 102603025-102603025 on Build GRCh38
rs765132384 Chr.X: 102602899-102602899 on Build GRCh38
rs777867447 Chr.X: 102602925-102602925 on Build GRCh38
rs746422685 Chr.X: 102602957-102602957 on Build GRCh38
rs763940953 Chr.X: 102603068-102603068 on Build GRCh38
rs371062313 Chr.X: 102602918-102602918 on Build GRCh38
rs749617854 Chr.X: 102602990-102602990 on Build GRCh38
rs1042668053 Chr.X: 102603003-102603003 on Build GRCh38
rs749534706 Chr.X: 102602985-102602985 on Build GRCh38
rs143305512 Chr.X: 102602940-102602940 on Build GRCh38
rs139067925 Chr.X: 102602970-102602970 on Build GRCh38
rs776025894 Chr.X: 102602984-102602984 on Build GRCh38
rs377359903 Chr.X: 102603058-102603058 on Build GRCh38
rs769008473 Chr.X: 102602996-102602996 on Build GRCh38
rs1056127986 Chr.X: 102602926-102602926 on Build GRCh38

View Full Product Details