Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184672272-184672497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIMPOL
Entrez Gene ID
201973
Gene Name
primase and DNA directed polymerase
Gene Aliases
CCDC111, MYP22
UniGene
Hs.481307
Gene Chromosome Location
Chr.4: 184649613-184694959 on Build GRCh38
Transcripts
NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447662 c.721G>T p.E241* Chr.4: 184672337-184672337 on Build GRCh38
5040905 c.728G>C p.W243S Chr.4: 184672344-184672344 on Build GRCh38
4381825 c.781C>A p.P261T Chr.4: 184672397-184672397 on Build GRCh38
6621311 c.844G>A p.G282R Chr.4: 184672460-184672460 on Build GRCh38
7314779 c.844+2_844+3insGGTCG p.? Chr.4: 184672462-184672463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762906128 Chr.4: 184672350-184672350 on Build GRCh38
rs776142895 Chr.4: 184672329-184672329 on Build GRCh38
rs780695010 Chr.4: 184672437-184672437 on Build GRCh38
rs374221499 Chr.4: 184672431-184672431 on Build GRCh38
rs775686581 Chr.4: 184672359-184672359 on Build GRCh38
rs746457375 Chr.4: 184672296-184672296 on Build GRCh38
rs768393267 Chr.4: 184672312-184672312 on Build GRCh38
rs771771068 Chr.4: 184672361-184672361 on Build GRCh38
rs749057543 Chr.4: 184672459-184672459 on Build GRCh38
rs760643017 Chr.4: 184672364-184672364 on Build GRCh38
rs774154627 Chr.4: 184672464-184672464 on Build GRCh38
rs750777798 Chr.4: 184672408-184672408 on Build GRCh38
rs1036846832 Chr.4: 184672402-184672402 on Build GRCh38
rs753996813 Chr.4: 184672376-184672376 on Build GRCh38
rs202179118 Chr.4: 184672416-184672416 on Build GRCh38
rs114174581 Chr.4: 184672377-184672377 on Build GRCh38
rs774998238 Chr.4: 184672339-184672339 on Build GRCh38
rs145554992 Chr.4: 184672434-184672434 on Build GRCh38
rs764446905 Chr.4: 184672371-184672371 on Build GRCh38
rs770893069 Chr.4: 184672460-184672460 on Build GRCh38
rs776711191 Chr.4: 184672478-184672478 on Build GRCh38
rs777270041 Chr.4: 184672454-184672454 on Build GRCh38
rs779743283 Chr.4: 184672293-184672293 on Build GRCh38
rs34772069 Chr.4: 184672397-184672397 on Build GRCh38
rs893003038 Chr.4: 184672368-184672368 on Build GRCh38
rs748032943 Chr.4: 184672331-184672331 on Build GRCh38
rs769392440 Chr.4: 184672451-184672451 on Build GRCh38
rs760869885 Chr.4: 184672468-184672468 on Build GRCh38
rs780686146 Chr.4: 184672420-184672420 on Build GRCh38
rs747759813 Chr.4: 184672450-184672450 on Build GRCh38
rs549819226 Chr.4: 184672351-184672351 on Build GRCh38
rs769589734 Chr.4: 184672337-184672337 on Build GRCh38
rs924429272 Chr.4: 184672332-184672332 on Build GRCh38

View Full Product Details