Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43995535-43995772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC24
Entrez Gene ID
149473
Gene Name
coiled-coil domain containing 24
Gene Aliases
-
UniGene
Hs.632394
Gene Chromosome Location
Chr.1: 43991247-43996528 on Build GRCh38
Transcripts
NM_152499.2 XM_017000423.1 XM_017000424.1 XM_017000425.1 XM_017000426.1 XM_017000427.1 XM_017000428.1 XM_017000429.1 XM_017000430.1 XM_017000431.1 XM_017000432.1 XM_017000433.1 XM_017000434.1 XM_017000435.1 XM_017000436.1 XM_017000437.1 XM_017000438.1 XM_017000439.1 XM_017000440.1 XM_017000441.1 XM_017000442.1 XM_017000445.1 XM_017000446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909950 c.554G>A p.R185H Chr.1: 43995602-43995602 on Build GRCh38
6366498 c.569A>G p.E190G Chr.1: 43995617-43995617 on Build GRCh38
909951 c.580C>T p.P194S Chr.1: 43995628-43995628 on Build GRCh38
1317527 c.599C>A p.A200E Chr.1: 43995647-43995647 on Build GRCh38
4669883 c.622+2T>C p.? Chr.1: 43995672-43995672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766259472 Chr.1: 43995608-43995608 on Build GRCh38
rs751532633 Chr.1: 43995613-43995613 on Build GRCh38
rs747531488 Chr.1: 43995657-43995657 on Build GRCh38
rs918477687 Chr.1: 43995723-43995723 on Build GRCh38
rs762060345 Chr.1: 43995671-43995671 on Build GRCh38
rs371879890 Chr.1: 43995564-43995564 on Build GRCh38
rs755769696 Chr.1: 43995727-43995727 on Build GRCh38
rs747511244 Chr.1: 43995747-43995747 on Build GRCh38
rs776071926 Chr.1: 43995654-43995654 on Build GRCh38
rs199609262 Chr.1: 43995659-43995659 on Build GRCh38
rs755790274 Chr.1: 43995634-43995634 on Build GRCh38
rs765423258 Chr.1: 43995675-43995675 on Build GRCh38
rs756685442 Chr.1: 43995615-43995616 on Build GRCh38
rs916405046 Chr.1: 43995628-43995628 on Build GRCh38
rs776853018 Chr.1: 43995594-43995594 on Build GRCh38
rs980925671 Chr.1: 43995557-43995557 on Build GRCh38
rs370432718 Chr.1: 43995602-43995602 on Build GRCh38
rs773448694 Chr.1: 43995680-43995680 on Build GRCh38
rs984628075 Chr.1: 43995607-43995607 on Build GRCh38
rs923190409 Chr.1: 43995650-43995650 on Build GRCh38
rs548289375 Chr.1: 43995699-43995699 on Build GRCh38
rs371444168 Chr.1: 43995601-43995601 on Build GRCh38
rs927742129 Chr.1: 43995593-43995593 on Build GRCh38
rs933273491 Chr.1: 43995656-43995656 on Build GRCh38
rs767342138 Chr.1: 43995719-43995719 on Build GRCh38
rs748758035 Chr.1: 43995641-43995641 on Build GRCh38
rs756767516 Chr.1: 43995730-43995730 on Build GRCh38
rs534776697 Chr.1: 43995720-43995720 on Build GRCh38
rs776925255 Chr.1: 43995666-43995666 on Build GRCh38
rs376686850 Chr.1: 43995627-43995627 on Build GRCh38
rs748952462 Chr.1: 43995728-43995728 on Build GRCh38
rs758313781 Chr.1: 43995604-43995604 on Build GRCh38
rs1053953580 Chr.1: 43995752-43995752 on Build GRCh38
rs777306784 Chr.1: 43995638-43995638 on Build GRCh38
rs145391003 Chr.1: 43995555-43995555 on Build GRCh38
rs550402568 Chr.1: 43995556-43995556 on Build GRCh38
rs778623280 Chr.1: 43995732-43995732 on Build GRCh38
rs367602623 Chr.1: 43995681-43995681 on Build GRCh38
rs776162111 Chr.1: 43995580-43995580 on Build GRCh38
rs761080442 Chr.1: 43995584-43995584 on Build GRCh38
rs376842836 Chr.1: 43995585-43995585 on Build GRCh38
rs754929963 Chr.1: 43995717-43995717 on Build GRCh38
rs751443066 Chr.1: 43995706-43995706 on Build GRCh38
rs756622233 Chr.1: 43995691-43995691 on Build GRCh38
rs754703867 Chr.1: 43995615-43995615 on Build GRCh38
rs780864246 Chr.1: 43995624-43995624 on Build GRCh38
rs766453640 Chr.1: 43995684-43995684 on Build GRCh38
rs761997453 Chr.1: 43995595-43995595 on Build GRCh38
rs552171928 Chr.1: 43995648-43995648 on Build GRCh38

View Full Product Details