Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145873663-145873422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD35
Entrez Gene ID
148741
Gene Name
ankyrin repeat domain 35
Gene Aliases
-
UniGene
Hs.710624
Gene Chromosome Location
Chr.1: 145866560-145885866 on Build GRCh38
Transcripts
NM_001280799.1 NM_144698.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3862589 c.1190C>T p.P397L Chr.1: 145873579-145873579 on Build GRCh38
7355677 c.1217A>T p.D406V Chr.1: 145873552-145873552 on Build GRCh38
895205 c.1345G>A p.A449T Chr.1: 145873424-145873424 on Build GRCh38
1638140 c.1246G>A p.V416I Chr.1: 145873523-145873523 on Build GRCh38
895204 c.1184T>C p.V395A Chr.1: 145873585-145873585 on Build GRCh38
View More

View Full Product Details