Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14704246-14704483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
-
UniGene
Hs.661452
Gene Chromosome Location
Chr.3: 14673532-14773036 on Build GRCh38
Transcripts
NM_001184957.1 NM_001184958.1 NM_032137.4 XM_006713341.2 XM_006713342.2 XM_006713343.2 XM_011534153.2 XM_011534154.2 XM_011534157.2 XM_011534158.2 XM_017007310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4402679 c.950A>C p.K317T Chr.3: 14704408-14704408 on Build GRCh38
3588472 c.961C>T p.H321Y Chr.3: 14704419-14704419 on Build GRCh38
1692416 c.904G>A p.G302R Chr.3: 14704362-14704362 on Build GRCh38
3588471 c.900G>A p.W300* Chr.3: 14704358-14704358 on Build GRCh38
1692415 c.890G>A p.R297K Chr.3: 14704348-14704348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570401629 Chr.3: 14704422-14704422 on Build GRCh38
rs764197817 Chr.3: 14704410-14704410 on Build GRCh38
rs758897746 Chr.3: 14704421-14704421 on Build GRCh38
rs199990486 Chr.3: 14704332-14704332 on Build GRCh38
rs777320067 Chr.3: 14704445-14704445 on Build GRCh38
rs148510620 Chr.3: 14704385-14704385 on Build GRCh38
rs776802278 Chr.3: 14704326-14704328 on Build GRCh38
rs751839189 Chr.3: 14704437-14704437 on Build GRCh38
rs755841263 Chr.3: 14704438-14704438 on Build GRCh38
rs146642906 Chr.3: 14704392-14704392 on Build GRCh38
rs770577830 Chr.3: 14704450-14704450 on Build GRCh38
rs778553841 Chr.3: 14704456-14704456 on Build GRCh38
rs748903637 Chr.3: 14704447-14704447 on Build GRCh38
rs774381028 Chr.3: 14704287-14704287 on Build GRCh38
rs780464445 Chr.3: 14704434-14704434 on Build GRCh38
rs17040196 Chr.3: 14704350-14704350 on Build GRCh38
rs762405197 Chr.3: 14704426-14704426 on Build GRCh38
rs759487310 Chr.3: 14704394-14704394 on Build GRCh38
rs780342083 Chr.3: 14704379-14704379 on Build GRCh38
rs759433853 Chr.3: 14704304-14704304 on Build GRCh38
rs760993564 Chr.3: 14704409-14704409 on Build GRCh38
rs753828172 Chr.3: 14704356-14704356 on Build GRCh38
rs756273724 Chr.3: 14704323-14704323 on Build GRCh38
rs114234710 Chr.3: 14704383-14704383 on Build GRCh38
rs768861528 Chr.3: 14704393-14704393 on Build GRCh38
rs111277329 Chr.3: 14704267-14704267 on Build GRCh38
rs775139083 Chr.3: 14704399-14704399 on Build GRCh38
rs752976445 Chr.3: 14704311-14704311 on Build GRCh38
rs369151755 Chr.3: 14704457-14704457 on Build GRCh38
rs757828816 Chr.3: 14704372-14704372 on Build GRCh38
rs546912466 Chr.3: 14704271-14704271 on Build GRCh38
rs758771146 Chr.3: 14704377-14704377 on Build GRCh38
rs772820995 Chr.3: 14704391-14704391 on Build GRCh38
rs765237727 Chr.3: 14704420-14704420 on Build GRCh38
rs772049661 Chr.3: 14704397-14704397 on Build GRCh38
rs767353891 Chr.3: 14704310-14704310 on Build GRCh38
rs761914883 Chr.3: 14704417-14704417 on Build GRCh38
rs1019815846 Chr.3: 14704322-14704322 on Build GRCh38
rs746336927 Chr.3: 14704373-14704373 on Build GRCh38

View Full Product Details