Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55270375-55270606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX29
Entrez Gene ID
54505
Gene Name
DEAH-box helicase 29
Gene Aliases
DDX29
UniGene
Hs.593268
Gene Chromosome Location
Chr.5: 55256245-55307693 on Build GRCh38
Transcripts
NM_019030.2 XM_006714653.2 XM_017009588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415184 c.3011A>T p.D1004V Chr.5: 55270470-55270470 on Build GRCh38
5637778 c.2975G>A p.R992Q Chr.5: 55270596-55270596 on Build GRCh38
7186348 c.3061C>G p.H1021D Chr.5: 55270420-55270420 on Build GRCh38
1068819 c.2971T>C p.F991L Chr.5: 55270600-55270600 on Build GRCh38
7206218 c.3046G>T p.E1016* Chr.5: 55270435-55270435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779803158 Chr.5: 55270446-55270446 on Build GRCh38
rs776983537 Chr.5: 55270527-55270527 on Build GRCh38
rs752776406 Chr.5: 55270549-55270549 on Build GRCh38
rs766483167 Chr.5: 55270513-55270513 on Build GRCh38
rs373491999 Chr.5: 55270404-55270404 on Build GRCh38
rs971424948 Chr.5: 55270548-55270548 on Build GRCh38
rs560990380 Chr.5: 55270474-55270474 on Build GRCh38
rs778043408 Chr.5: 55270427-55270427 on Build GRCh38
rs769280893 Chr.5: 55270442-55270442 on Build GRCh38
rs773095186 Chr.5: 55270494-55270494 on Build GRCh38
rs886268166 Chr.5: 55270506-55270506 on Build GRCh38
rs370774198 Chr.5: 55270571-55270571 on Build GRCh38
rs868769705 Chr.5: 55270577-55270577 on Build GRCh38
rs191521787 Chr.5: 55270402-55270402 on Build GRCh38
rs747403295 Chr.5: 55270430-55270430 on Build GRCh38
rs184086503 Chr.5: 55270539-55270539 on Build GRCh38
rs888937584 Chr.5: 55270417-55270417 on Build GRCh38
rs760599480 Chr.5: 55270505-55270505 on Build GRCh38
rs111401562 Chr.5: 55270575-55270575 on Build GRCh38
rs748716619 Chr.5: 55270447-55270447 on Build GRCh38
rs753646587 Chr.5: 55270559-55270559 on Build GRCh38
rs371480219 Chr.5: 55270484-55270484 on Build GRCh38
rs768278824 Chr.5: 55270462-55270462 on Build GRCh38
rs758860943 Chr.5: 55270553-55270553 on Build GRCh38
rs376935780 Chr.5: 55270547-55270547 on Build GRCh38
rs766731469 Chr.5: 55270541-55270541 on Build GRCh38
rs764605356 Chr.5: 55270556-55270556 on Build GRCh38
rs376975893 Chr.5: 55270414-55270414 on Build GRCh38
rs1035908293 Chr.5: 55270519-55270519 on Build GRCh38
rs1027511485 Chr.5: 55270451-55270451 on Build GRCh38
rs150568756 Chr.5: 55270418-55270418 on Build GRCh38

View Full Product Details