Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10793671-10793944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM2
Entrez Gene ID
1785
Gene Name
dynamin 2
Gene Aliases
CMT2M, CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII, LCCS5
UniGene
Hs.211463
Gene Chromosome Location
Chr.19: 10718053-10831910 on Build GRCh38
Transcripts
NM_001005360.2 NM_001005361.2 NM_001005362.2 NM_001190716.1 NM_004945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250478 c.1082G>T p.R361L Chr.19: 10793809-10793809 on Build GRCh38
4678840 c.1006delT p.F336fs*40 Chr.19: 10793733-10793733 on Build GRCh38
2813176 c.993-1G>A p.? Chr.19: 10793719-10793719 on Build GRCh38
6999039 c.1078G>A p.A360T Chr.19: 10793805-10793805 on Build GRCh38
4167823 c.1002G>T p.Q334H Chr.19: 10793729-10793729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779676389 Chr.19: 10793861-10793862 on Build GRCh38
rs121909092 Chr.19: 10793829-10793829 on Build GRCh38
rs750577663 Chr.19: 10793801-10793801 on Build GRCh38
rs537132157 Chr.19: 10793867-10793867 on Build GRCh38
rs753981065 Chr.19: 10793772-10793772 on Build GRCh38
rs761380478 Chr.19: 10793706-10793706 on Build GRCh38
rs746959606 Chr.19: 10793822-10793822 on Build GRCh38
rs776227722 Chr.19: 10793704-10793704 on Build GRCh38
rs775764080 Chr.19: 10793871-10793871 on Build GRCh38
rs781330890 Chr.19: 10793878-10793878 on Build GRCh38
rs876661176 Chr.19: 10793778-10793778 on Build GRCh38
rs549033523 Chr.19: 10793729-10793729 on Build GRCh38
rs772959277 Chr.19: 10793862-10793862 on Build GRCh38
rs767838513 Chr.19: 10793737-10793737 on Build GRCh38
rs747050783 Chr.19: 10793759-10793759 on Build GRCh38
rs753224039 Chr.19: 10793739-10793739 on Build GRCh38
rs750682509 Chr.19: 10793897-10793897 on Build GRCh38
rs769166411 Chr.19: 10793861-10793861 on Build GRCh38
rs185654406 Chr.19: 10793883-10793883 on Build GRCh38
rs864309705 Chr.19: 10793748-10793748 on Build GRCh38
rs772437963 Chr.19: 10793868-10793868 on Build GRCh38
rs112238216 Chr.19: 10793804-10793804 on Build GRCh38
rs759996048 Chr.19: 10793731-10793731 on Build GRCh38
rs754274733 Chr.19: 10793879-10793879 on Build GRCh38
rs769556504 Chr.19: 10793721-10793721 on Build GRCh38
rs758636844 Chr.19: 10793906-10793906 on Build GRCh38
rs375069966 Chr.19: 10793699-10793699 on Build GRCh38
rs757266860 Chr.19: 10793777-10793777 on Build GRCh38
rs121909089 Chr.19: 10793833-10793833 on Build GRCh38
rs764163643 Chr.19: 10793768-10793768 on Build GRCh38
rs372507531 Chr.19: 10793877-10793877 on Build GRCh38
rs267606772 Chr.19: 10793799-10793799 on Build GRCh38
rs121909090 Chr.19: 10793832-10793832 on Build GRCh38
rs747752571 Chr.19: 10793831-10793831 on Build GRCh38
rs201700534 Chr.19: 10793837-10793837 on Build GRCh38
rs779949390 Chr.19: 10793814-10793814 on Build GRCh38
rs587783594 Chr.19: 10793851-10793851 on Build GRCh38
rs768651920 Chr.19: 10793828-10793828 on Build GRCh38
rs367698794 Chr.19: 10793884-10793884 on Build GRCh38
rs779241319 Chr.19: 10793798-10793798 on Build GRCh38
rs1010041153 Chr.19: 10793865-10793865 on Build GRCh38

View Full Product Details