Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69332679-69332910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG8
Entrez Gene ID
84342
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
CDG2H, DOR1
UniGene
Hs.130849
Gene Chromosome Location
Chr.16: 69328621-69339623 on Build GRCh38
Transcripts
NM_032382.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972874 c.1414-1G>T p.? Chr.16: 69332883-69332883 on Build GRCh38
6632275 c.1535G>A p.R512H Chr.16: 69332761-69332761 on Build GRCh38
5820613 c.1498A>T p.T500S Chr.16: 69332798-69332798 on Build GRCh38
1749727 c.1513G>A p.D505N Chr.16: 69332783-69332783 on Build GRCh38
6632273 c.1561G>A p.A521T Chr.16: 69332735-69332735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147051782 Chr.16: 69332713-69332713 on Build GRCh38
rs367923608 Chr.16: 69332852-69332852 on Build GRCh38
rs533567997 Chr.16: 69332758-69332758 on Build GRCh38
rs376659578 Chr.16: 69332772-69332772 on Build GRCh38
rs202134146 Chr.16: 69332828-69332828 on Build GRCh38
rs890271209 Chr.16: 69332716-69332716 on Build GRCh38
rs751469714 Chr.16: 69332736-69332736 on Build GRCh38
rs771239579 Chr.16: 69332793-69332793 on Build GRCh38
rs761841941 Chr.16: 69332827-69332827 on Build GRCh38
rs889540830 Chr.16: 69332710-69332710 on Build GRCh38
rs138398268 Chr.16: 69332747-69332747 on Build GRCh38
rs147250033 Chr.16: 69332805-69332805 on Build GRCh38
rs755033921 Chr.16: 69332728-69332728 on Build GRCh38
rs760985243 Chr.16: 69332714-69332714 on Build GRCh38
rs769286853 Chr.16: 69332726-69332729 on Build GRCh38
rs752727817 Chr.16: 69332823-69332823 on Build GRCh38
rs766441556 Chr.16: 69332715-69332715 on Build GRCh38
rs779892154 Chr.16: 69332846-69332846 on Build GRCh38
rs143808112 Chr.16: 69332762-69332762 on Build GRCh38
rs764087834 Chr.16: 69332732-69332732 on Build GRCh38
rs773626119 Chr.16: 69332781-69332781 on Build GRCh38
rs759654404 Chr.16: 69332808-69332808 on Build GRCh38
rs778596903 Chr.16: 69332855-69332855 on Build GRCh38
rs776832489 Chr.16: 69332802-69332802 on Build GRCh38
rs138741747 Chr.16: 69332829-69332829 on Build GRCh38
rs1032712624 Chr.16: 69332740-69332740 on Build GRCh38
rs760887437 Chr.16: 69332790-69332790 on Build GRCh38
rs768247840 Chr.16: 69332769-69332769 on Build GRCh38
rs753925473 Chr.16: 69332721-69332721 on Build GRCh38
rs3027 Chr.16: 69332746-69332746 on Build GRCh38
rs187701280 Chr.16: 69332737-69332737 on Build GRCh38
rs200031891 Chr.16: 69332821-69332821 on Build GRCh38
rs781067037 Chr.16: 69332744-69332744 on Build GRCh38
rs541323524 Chr.16: 69332742-69332742 on Build GRCh38
rs368121830 Chr.16: 69332711-69332711 on Build GRCh38
rs1010178053 Chr.16: 69332750-69332750 on Build GRCh38
rs533170895 Chr.16: 69332773-69332773 on Build GRCh38
rs113368039 Chr.16: 69332853-69332853 on Build GRCh38
rs755798031 Chr.16: 69332754-69332754 on Build GRCh38
rs200963000 Chr.16: 69332890-69332890 on Build GRCh38

View Full Product Details