Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 33867282-33867543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVEN
Entrez Gene ID
57099
Gene Name
apoptosis and caspase activation inhibitor
Gene Aliases
PDCD12
UniGene
Hs.555966
Gene Chromosome Location
Chr.15: 33858602-34074877 on Build GRCh38
Transcripts
NM_020371.2 XM_011521818.2 XM_011521819.2 XM_011521820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
13994 c.937T>C p.S313P Chr.15: 33867531-33867531 on Build GRCh38
960764 c.962A>G p.Q321R Chr.15: 33867506-33867506 on Build GRCh38
459055 c.932T>C p.L311P Chr.15: 33867536-33867536 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775431878 Chr.15: 33867492-33867492 on Build GRCh38
rs183071578 Chr.15: 33867332-33867332 on Build GRCh38
rs185248315 Chr.15: 33867430-33867430 on Build GRCh38
rs1041995469 Chr.15: 33867489-33867489 on Build GRCh38
rs747263167 Chr.15: 33867456-33867457 on Build GRCh38
rs570842145 Chr.15: 33867472-33867472 on Build GRCh38
rs990049928 Chr.15: 33867434-33867434 on Build GRCh38
rs958145815 Chr.15: 33867509-33867509 on Build GRCh38
rs750954353 Chr.15: 33867511-33867511 on Build GRCh38
rs754521315 Chr.15: 33867465-33867465 on Build GRCh38
rs751418468 Chr.15: 33867308-33867308 on Build GRCh38
rs780510838 Chr.15: 33867466-33867466 on Build GRCh38
rs568981138 Chr.15: 33867318-33867318 on Build GRCh38
rs774372596 Chr.15: 33867480-33867480 on Build GRCh38
rs142846842 Chr.15: 33867303-33867303 on Build GRCh38
rs373416760 Chr.15: 33867481-33867481 on Build GRCh38
rs544232874 Chr.15: 33867408-33867408 on Build GRCh38
rs199852977 Chr.15: 33867419-33867419 on Build GRCh38
rs903021846 Chr.15: 33867495-33867495 on Build GRCh38
rs142129057 Chr.15: 33867337-33867337 on Build GRCh38
rs532432476 Chr.15: 33867302-33867302 on Build GRCh38
rs749066672 Chr.15: 33867475-33867475 on Build GRCh38
rs764282186 Chr.15: 33867499-33867499 on Build GRCh38
rs1032845259 Chr.15: 33867503-33867503 on Build GRCh38
rs780210423 Chr.15: 33867446-33867447 on Build GRCh38
rs755823808 Chr.15: 33867471-33867471 on Build GRCh38
rs372221789 Chr.15: 33867502-33867502 on Build GRCh38
rs781120619 Chr.15: 33867327-33867327 on Build GRCh38
rs763355655 Chr.15: 33867514-33867514 on Build GRCh38
rs533975083 Chr.15: 33867349-33867349 on Build GRCh38
rs750856935 Chr.15: 33867458-33867458 on Build GRCh38
rs959806801 Chr.15: 33867352-33867352 on Build GRCh38
rs770798507 Chr.15: 33867478-33867478 on Build GRCh38
rs760999721 Chr.15: 33867494-33867494 on Build GRCh38
rs757503602 Chr.15: 33867341-33867341 on Build GRCh38
rs768937925 Chr.15: 33867492-33867492 on Build GRCh38
rs1007763392 Chr.15: 33867398-33867398 on Build GRCh38
rs193037128 Chr.15: 33867337-33867337 on Build GRCh38
rs768810356 Chr.15: 33867359-33867359 on Build GRCh38
rs111712854 Chr.15: 33867333-33867333 on Build GRCh38
rs965714558 Chr.15: 33867415-33867415 on Build GRCh38
rs747891598 Chr.15: 33867467-33867467 on Build GRCh38
rs978212468 Chr.15: 33867336-33867336 on Build GRCh38
rs1033973979 Chr.15: 33867304-33867304 on Build GRCh38
rs374881660 Chr.15: 33867429-33867429 on Build GRCh38
rs765518611 Chr.15: 33867508-33867508 on Build GRCh38
rs975734709 Chr.15: 33867418-33867418 on Build GRCh38
rs772255503 Chr.15: 33867487-33867487 on Build GRCh38
rs776787283 Chr.15: 33867501-33867501 on Build GRCh38
rs954227324 Chr.15: 33867395-33867395 on Build GRCh38

View Full Product Details