Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63323823-63324070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD3
Entrez Gene ID
27022
Gene Name
forkhead box D3
Gene Aliases
AIS1, Genesis, HFH2, VAMAS2
UniGene
Hs.546573
Gene Chromosome Location
Chr.1: 63323059-63325126 on Build GRCh38
Transcripts
NM_012183.2 NR_121637.1

Gene Symbol
FOXD3-AS1
Entrez Gene ID
100996301
Gene Name
FOXD3 antisense RNA 1 (head to head)
Gene Aliases
pasFOXD3
UniGene
Gene Chromosome Location
Chr.1: 63320884-63324441 on Build GRCh38
Transcripts
NM_012183.2 NR_121637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3377256 c.769G>A p.G257S Chr.1: 63323827-63323827 on Build GRCh38
6435746 c.787G>A p.A263T Chr.1: 63323845-63323845 on Build GRCh38
6318921 c.793G>T p.A265S Chr.1: 63323851-63323851 on Build GRCh38
536016 c.806G>T p.G269V Chr.1: 63323864-63323864 on Build GRCh38
6540489 c.1001G>T p.G334V Chr.1: 63324059-63324059 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034849768 Chr.1: 63324040-63324040 on Build GRCh38
rs781209290 Chr.1: 63323878-63323878 on Build GRCh38
rs896298016 Chr.1: 63324051-63324051 on Build GRCh38
rs764763503 Chr.1: 63323860-63323860 on Build GRCh38
rs541120234 Chr.1: 63323908-63323908 on Build GRCh38
rs765929377 Chr.1: 63323854-63323854 on Build GRCh38
rs1024977459 Chr.1: 63323840-63323840 on Build GRCh38
rs769912149 Chr.1: 63323884-63323884 on Build GRCh38
rs766446421 Chr.1: 63323911-63323911 on Build GRCh38
rs923547339 Chr.1: 63323892-63323892 on Build GRCh38
rs758688806 Chr.1: 63323862-63323862 on Build GRCh38
rs760588882 Chr.1: 63323888-63323888 on Build GRCh38
rs754633262 Chr.1: 63323857-63323857 on Build GRCh38
rs759091658 Chr.1: 63324039-63324039 on Build GRCh38
rs1047985255 Chr.1: 63323895-63323895 on Build GRCh38
rs910628448 Chr.1: 63323876-63323876 on Build GRCh38
rs745916627 Chr.1: 63323879-63323879 on Build GRCh38
rs940972560 Chr.1: 63323952-63323952 on Build GRCh38
rs771695051 Chr.1: 63323898-63323898 on Build GRCh38
rs904940701 Chr.1: 63324022-63324022 on Build GRCh38
rs979931976 Chr.1: 63323875-63323875 on Build GRCh38
rs1038563736 Chr.1: 63323987-63323987 on Build GRCh38
rs773073356 Chr.1: 63323900-63323900 on Build GRCh38
rs774979471 Chr.1: 63323871-63323871 on Build GRCh38
rs970294009 Chr.1: 63323967-63323967 on Build GRCh38
rs776745749 Chr.1: 63324038-63324038 on Build GRCh38
rs757786681 Chr.1: 63323874-63323874 on Build GRCh38
rs747461169 Chr.1: 63323864-63323864 on Build GRCh38
rs752227625 Chr.1: 63324052-63324052 on Build GRCh38
rs753420481 Chr.1: 63323855-63323855 on Build GRCh38
rs752440666 Chr.1: 63323861-63323861 on Build GRCh38
rs532147202 Chr.1: 63323865-63323866 on Build GRCh38
rs749589976 Chr.1: 63323897-63323897 on Build GRCh38
rs868477983 Chr.1: 63323931-63323931 on Build GRCh38
rs764858096 Chr.1: 63324050-63324050 on Build GRCh38
rs775562720 Chr.1: 63323891-63323891 on Build GRCh38
rs1002472593 Chr.1: 63324033-63324033 on Build GRCh38

View Full Product Details