Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74655648-74655907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
UniGene
Hs.682454
Gene Chromosome Location
Chr.15: 74630558-74696045 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3 XM_011522092.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
434298 c.664A>G p.I222V Chr.15: 74655889-74655889 on Build GRCh38
6329330 c.775C>T p.R259* Chr.15: 74655778-74655778 on Build GRCh38
1749309 c.742G>T p.E248* Chr.15: 74655811-74655811 on Build GRCh38
4056905 c.759C>T p.S253S Chr.15: 74655794-74655794 on Build GRCh38
4056906 c.710C>G p.P237R Chr.15: 74655843-74655843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752566564 Chr.15: 74655880-74655880 on Build GRCh38
rs772170307 Chr.15: 74655777-74655777 on Build GRCh38
rs971982773 Chr.15: 74655850-74655850 on Build GRCh38
rs561328785 Chr.15: 74655794-74655794 on Build GRCh38
rs556581951 Chr.15: 74655709-74655709 on Build GRCh38
rs7497342 Chr.15: 74655722-74655722 on Build GRCh38
rs775891360 Chr.15: 74655825-74655825 on Build GRCh38
rs932039438 Chr.15: 74655676-74655676 on Build GRCh38
rs140728378 Chr.15: 74655787-74655787 on Build GRCh38
rs960566889 Chr.15: 74655872-74655872 on Build GRCh38
rs776787766 Chr.15: 74655793-74655793 on Build GRCh38
rs953361566 Chr.15: 74655757-74655757 on Build GRCh38
rs377491253 Chr.15: 74655879-74655879 on Build GRCh38
rs755570696 Chr.15: 74655760-74655760 on Build GRCh38
rs772333439 Chr.15: 74655708-74655708 on Build GRCh38
rs767052992 Chr.15: 74655756-74655756 on Build GRCh38
rs892679695 Chr.15: 74655735-74655735 on Build GRCh38
rs760801853 Chr.15: 74655718-74655718 on Build GRCh38
rs759824978 Chr.15: 74655747-74655747 on Build GRCh38
rs75726871 Chr.15: 74655690-74655690 on Build GRCh38
rs76943949 Chr.15: 74655734-74655734 on Build GRCh38
rs748142645 Chr.15: 74655775-74655775 on Build GRCh38
rs755521800 Chr.15: 74655753-74655753 on Build GRCh38
rs147487761 Chr.15: 74655754-74655754 on Build GRCh38
rs754633529 Chr.15: 74655878-74655878 on Build GRCh38
rs763263440 Chr.15: 74655852-74655852 on Build GRCh38
rs145840933 Chr.15: 74655769-74655769 on Build GRCh38
rs765905853 Chr.15: 74655860-74655860 on Build GRCh38
rs369652432 Chr.15: 74655755-74655755 on Build GRCh38
rs779584456 Chr.15: 74655763-74655763 on Build GRCh38
rs770978061 Chr.15: 74655791-74655791 on Build GRCh38
rs1034873520 Chr.15: 74655855-74655855 on Build GRCh38
rs920437460 Chr.15: 74655861-74655861 on Build GRCh38
rs189497300 Chr.15: 74655859-74655859 on Build GRCh38
rs764782564 Chr.15: 74655716-74655716 on Build GRCh38
rs538490167 Chr.15: 74655696-74655696 on Build GRCh38
rs752378157 Chr.15: 74655708-74655708 on Build GRCh38
rs145915480 Chr.15: 74655815-74655815 on Build GRCh38
rs778751798 Chr.15: 74655774-74655774 on Build GRCh38
rs777808860 Chr.15: 74655778-74655778 on Build GRCh38
rs1001939113 Chr.15: 74655851-74655851 on Build GRCh38
rs766985395 Chr.15: 74655686-74655689 on Build GRCh38
rs776240001 Chr.15: 74655702-74655702 on Build GRCh38
rs753487087 Chr.15: 74655862-74655862 on Build GRCh38
rs753460829 Chr.15: 74655768-74655768 on Build GRCh38
rs1009603616 Chr.15: 74655723-74655723 on Build GRCh38
rs764605310 Chr.15: 74655853-74655853 on Build GRCh38
rs748690391 Chr.15: 74655785-74655785 on Build GRCh38

View Full Product Details