Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73241338-73241573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
UniGene
Hs.433062
Gene Chromosome Location
Chr.6: 73241314-73310365 on Build GRCh38
Transcripts
NM_001251874.1 NM_030568.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4397597 c.464C>T p.S155L Chr.6: 73241560-73241560 on Build GRCh38
4856205 c.490C>T p.P164S Chr.6: 73241534-73241534 on Build GRCh38
3629640 c.491C>T p.P164L Chr.6: 73241533-73241533 on Build GRCh38
4586915 c.453G>A p.V151V Chr.6: 73241571-73241571 on Build GRCh38
4572220 c.459T>C p.G153G Chr.6: 73241565-73241565 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765427624 Chr.6: 73241514-73241514 on Build GRCh38
rs562732290 Chr.6: 73241433-73241433 on Build GRCh38
rs772596344 Chr.6: 73241483-73241483 on Build GRCh38
rs1034454909 Chr.6: 73241422-73241422 on Build GRCh38
rs1012597067 Chr.6: 73241510-73241510 on Build GRCh38
rs756908211 Chr.6: 73241550-73241550 on Build GRCh38
rs747755706 Chr.6: 73241400-73241400 on Build GRCh38
rs117702866 Chr.6: 73241401-73241401 on Build GRCh38
rs941488701 Chr.6: 73241425-73241425 on Build GRCh38
rs538554497 Chr.6: 73241533-73241533 on Build GRCh38
rs376634472 Chr.6: 73241502-73241502 on Build GRCh38
rs530066779 Chr.6: 73241434-73241434 on Build GRCh38
rs1014849992 Chr.6: 73241458-73241458 on Build GRCh38
rs73757029 Chr.6: 73241460-73241460 on Build GRCh38
rs373479803 Chr.6: 73241497-73241497 on Build GRCh38
rs776040074 Chr.6: 73241489-73241489 on Build GRCh38
rs143460400 Chr.6: 73241522-73241522 on Build GRCh38
rs778100429 Chr.6: 73241545-73241545 on Build GRCh38
rs779715428 Chr.6: 73241423-73241423 on Build GRCh38
rs758714404 Chr.6: 73241518-73241518 on Build GRCh38
rs939603983 Chr.6: 73241453-73241453 on Build GRCh38
rs182325320 Chr.6: 73241532-73241532 on Build GRCh38
rs932691641 Chr.6: 73241398-73241398 on Build GRCh38
rs532829513 Chr.6: 73241397-73241397 on Build GRCh38
rs566208834 Chr.6: 73241441-73241441 on Build GRCh38
rs976698998 Chr.6: 73241370-73241370 on Build GRCh38
rs528287481 Chr.6: 73241511-73241511 on Build GRCh38
rs753386664 Chr.6: 73241542-73241542 on Build GRCh38
rs895459650 Chr.6: 73241470-73241470 on Build GRCh38
rs767550112 Chr.6: 73241523-73241523 on Build GRCh38
rs750381929 Chr.6: 73241389-73241389 on Build GRCh38

View Full Product Details