Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121872554-121872825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF2
Entrez Gene ID
55840
Gene Name
ELL associated factor 2
Gene Aliases
BM040, TRAITS, U19
UniGene
Hs.477325
Gene Chromosome Location
Chr.3: 121835180-121886526 on Build GRCh38
Transcripts
NM_001320041.1 NM_018456.5 XM_005247618.3 XM_017006861.1 XM_017006862.1 XM_017006863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1693277 c.674C>T p.P225L Chr.3: 121872726-121872726 on Build GRCh38
7365789 c.708C>G p.D236E Chr.3: 121872760-121872760 on Build GRCh38
4554701 c.628G>A p.G210R Chr.3: 121872680-121872680 on Build GRCh38
479291 c.612C>A p.S204S Chr.3: 121872664-121872664 on Build GRCh38
5011178 c.518T>G p.M173R Chr.3: 121872570-121872570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375955486 Chr.3: 121872682-121872682 on Build GRCh38
rs148765156 Chr.3: 121872748-121872748 on Build GRCh38
rs200859609 Chr.3: 121872672-121872672 on Build GRCh38
rs754344006 Chr.3: 121872721-121872721 on Build GRCh38
rs372997631 Chr.3: 121872715-121872715 on Build GRCh38
rs762151439 Chr.3: 121872702-121872702 on Build GRCh38
rs541758349 Chr.3: 121872736-121872736 on Build GRCh38
rs773466387 Chr.3: 121872785-121872785 on Build GRCh38
rs770777296 Chr.3: 121872691-121872691 on Build GRCh38
rs775387376 Chr.3: 121872701-121872701 on Build GRCh38
rs144356897 Chr.3: 121872594-121872594 on Build GRCh38
rs759371140 Chr.3: 121872602-121872602 on Build GRCh38
rs937449936 Chr.3: 121872709-121872709 on Build GRCh38
rs139363943 Chr.3: 121872760-121872760 on Build GRCh38
rs115221306 Chr.3: 121872769-121872769 on Build GRCh38
rs779689447 Chr.3: 121872791-121872791 on Build GRCh38
rs756218295 Chr.3: 121872714-121872714 on Build GRCh38
rs759299979 Chr.3: 121872699-121872699 on Build GRCh38
rs777339809 Chr.3: 121872742-121872742 on Build GRCh38
rs780547951 Chr.3: 121872655-121872655 on Build GRCh38
rs745851836 Chr.3: 121872756-121872756 on Build GRCh38
rs760559147 Chr.3: 121872593-121872593 on Build GRCh38
rs767793472 Chr.3: 121872710-121872710 on Build GRCh38
rs199994792 Chr.3: 121872637-121872637 on Build GRCh38
rs370593056 Chr.3: 121872755-121872755 on Build GRCh38
rs376558625 Chr.3: 121872754-121872754 on Build GRCh38
rs556047695 Chr.3: 121872621-121872621 on Build GRCh38
rs773171966 Chr.3: 121872585-121872585 on Build GRCh38
rs369018167 Chr.3: 121872694-121872694 on Build GRCh38
rs913810218 Chr.3: 121872588-121872588 on Build GRCh38
rs750485158 Chr.3: 121872652-121872652 on Build GRCh38
rs766289522 Chr.3: 121872597-121872597 on Build GRCh38
rs771670026 Chr.3: 121872674-121872674 on Build GRCh38
rs778582989 Chr.3: 121872669-121872669 on Build GRCh38
rs769047516 Chr.3: 121872665-121872665 on Build GRCh38
rs758058371 Chr.3: 121872740-121872740 on Build GRCh38
rs748460863 Chr.3: 121872738-121872738 on Build GRCh38
rs747885285 Chr.3: 121872781-121872781 on Build GRCh38
rs760173706 Chr.3: 121872685-121872685 on Build GRCh38
rs767640852 Chr.3: 121872786-121872789 on Build GRCh38
rs765094507 Chr.3: 121872700-121872700 on Build GRCh38
rs764281821 Chr.3: 121872615-121872615 on Build GRCh38
rs781456291 Chr.3: 121872627-121872627 on Build GRCh38
rs753587719 Chr.3: 121872599-121872599 on Build GRCh38
rs750477617 Chr.3: 121872712-121872712 on Build GRCh38
rs759619071 Chr.3: 121872595-121872595 on Build GRCh38
rs762774106 Chr.3: 121872764-121872764 on Build GRCh38
rs757455230 Chr.3: 121872623-121872623 on Build GRCh38
rs775442349 Chr.3: 121872762-121872762 on Build GRCh38
rs749678234 Chr.3: 121872664-121872664 on Build GRCh38

View Full Product Details