Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244842922-244843161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX20
Entrez Gene ID
116228
Gene Name
COX20, cytochrome c oxidase assembly factor
Gene Aliases
FAM36A
UniGene
Hs.411490
Gene Chromosome Location
Chr.1: 244835306-244845063 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_001312871.1 NM_001312872.1 NM_001312873.1 NM_001312874.1 NM_198076.5 NR_132419.1 NR_132420.1 NR_132421.1 XM_017001115.1 XM_017001116.1 XM_017001117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641715 c.258-2A>G p.? Chr.1: 244843039-244843039 on Build GRCh38
3486407 c.307G>A p.G103S Chr.1: 244843126-244843126 on Build GRCh38
7185550 c.288A>G p.K96K Chr.1: 244843107-244843107 on Build GRCh38
4748233 c.332_334delTAT p.L112delL Chr.1: 244843115-244843117 on Build GRCh38
2051454 c.266G>T p.R89I Chr.1: 244843085-244843085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780760696 Chr.1: 244843047-244843047 on Build GRCh38
rs200065889 Chr.1: 244843038-244843038 on Build GRCh38
rs397829223 Chr.1: 244843067-244843067 on Build GRCh38
rs959962433 Chr.1: 244842972-244842972 on Build GRCh38
rs749626450 Chr.1: 244843077-244843077 on Build GRCh38
rs1053697 Chr.1: 244843137-244843137 on Build GRCh38
rs777326199 Chr.1: 244843076-244843076 on Build GRCh38
rs988125986 Chr.1: 244843114-244843114 on Build GRCh38
rs889035347 Chr.1: 244843118-244843118 on Build GRCh38
rs750263153 Chr.1: 244843022-244843022 on Build GRCh38
rs769116815 Chr.1: 244843138-244843138 on Build GRCh38
rs868648645 Chr.1: 244843033-244843033 on Build GRCh38
rs776767802 Chr.1: 244843120-244843120 on Build GRCh38
rs747702163 Chr.1: 244843053-244843053 on Build GRCh38
rs969459231 Chr.1: 244843052-244843052 on Build GRCh38
rs764346782 Chr.1: 244843113-244843113 on Build GRCh38
rs766346210 Chr.1: 244843131-244843131 on Build GRCh38
rs771647113 Chr.1: 244843090-244843090 on Build GRCh38
rs1023227356 Chr.1: 244843025-244843025 on Build GRCh38
rs778454364 Chr.1: 244843015-244843018 on Build GRCh38
rs775660475 Chr.1: 244843094-244843094 on Build GRCh38
rs767647989 Chr.1: 244842997-244842997 on Build GRCh38
rs150172035 Chr.1: 244843088-244843088 on Build GRCh38
rs923502888 Chr.1: 244843117-244843117 on Build GRCh38
rs751606534 Chr.1: 244843046-244843046 on Build GRCh38
rs771987123 Chr.1: 244843093-244843093 on Build GRCh38
rs759911217 Chr.1: 244843105-244843105 on Build GRCh38
rs771340542 Chr.1: 244843081-244843081 on Build GRCh38
rs373502901 Chr.1: 244843138-244843138 on Build GRCh38
rs11544344 Chr.1: 244843127-244843127 on Build GRCh38
rs764892273 Chr.1: 244843021-244843021 on Build GRCh38
rs863223946 Chr.1: 244843045-244843045 on Build GRCh38
rs925425021 Chr.1: 244843068-244843068 on Build GRCh38
rs766397584 Chr.1: 244843041-244843041 on Build GRCh38
rs779792502 Chr.1: 244843108-244843110 on Build GRCh38
rs145325896 Chr.1: 244843135-244843135 on Build GRCh38
rs371098560 Chr.1: 244843128-244843128 on Build GRCh38
rs774833234 Chr.1: 244843086-244843086 on Build GRCh38
rs892277959 Chr.1: 244843015-244843015 on Build GRCh38
rs78677269 Chr.1: 244843005-244843005 on Build GRCh38
rs935322065 Chr.1: 244843085-244843085 on Build GRCh38
rs755816815 Chr.1: 244843055-244843055 on Build GRCh38
rs747291859 Chr.1: 244843115-244843117 on Build GRCh38
rs760963966 Chr.1: 244843013-244843013 on Build GRCh38
rs745397579 Chr.1: 244843026-244843030 on Build GRCh38
rs751479035 Chr.1: 244843133-244843133 on Build GRCh38

View Full Product Details