Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19915252-19915525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005742.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204997 c.1281T>G p.Y427* Chr.3: 19915310-19915310 on Build GRCh38
273581 c.1234G>T p.E412* Chr.3: 19915357-19915357 on Build GRCh38
7168147 c.1230T>G p.F410L Chr.3: 19915361-19915361 on Build GRCh38
730429 c.1233A>G p.K411K Chr.3: 19915358-19915358 on Build GRCh38
6876669 c.1215C>T p.S405S Chr.3: 19915376-19915376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755470847 Chr.3: 19915409-19915409 on Build GRCh38
rs769970752 Chr.3: 19915331-19915331 on Build GRCh38
rs73188472 Chr.3: 19915423-19915423 on Build GRCh38
rs753414107 Chr.3: 19915278-19915278 on Build GRCh38
rs758849936 Chr.3: 19915290-19915290 on Build GRCh38
rs139478971 Chr.3: 19915309-19915309 on Build GRCh38
rs757435535 Chr.3: 19915458-19915458 on Build GRCh38
rs543473770 Chr.3: 19915496-19915496 on Build GRCh38
rs915486758 Chr.3: 19915420-19915420 on Build GRCh38
rs771860890 Chr.3: 19915340-19915340 on Build GRCh38
rs761546608 Chr.3: 19915283-19915283 on Build GRCh38
rs887240860 Chr.3: 19915505-19915505 on Build GRCh38
rs768774634 Chr.3: 19915363-19915363 on Build GRCh38
rs972721426 Chr.3: 19915487-19915487 on Build GRCh38
rs546854889 Chr.3: 19915304-19915304 on Build GRCh38
rs906543980 Chr.3: 19915471-19915471 on Build GRCh38
rs766706351 Chr.3: 19915398-19915398 on Build GRCh38
rs764778068 Chr.3: 19915386-19915386 on Build GRCh38
rs201005662 Chr.3: 19915394-19915394 on Build GRCh38
rs755487283 Chr.3: 19915304-19915304 on Build GRCh38
rs749461148 Chr.3: 19915448-19915448 on Build GRCh38
rs756169185 Chr.3: 19915425-19915425 on Build GRCh38
rs762512808 Chr.3: 19915395-19915395 on Build GRCh38
rs776279101 Chr.3: 19915368-19915368 on Build GRCh38
rs750726180 Chr.3: 19915289-19915289 on Build GRCh38
rs777746825 Chr.3: 19915427-19915427 on Build GRCh38
rs778105687 Chr.3: 19915332-19915332 on Build GRCh38
rs765009835 Chr.3: 19915287-19915287 on Build GRCh38
rs965359828 Chr.3: 19915475-19915475 on Build GRCh38
rs1042532993 Chr.3: 19915366-19915366 on Build GRCh38
rs780310273 Chr.3: 19915300-19915300 on Build GRCh38
rs752751858 Chr.3: 19915424-19915424 on Build GRCh38
rs748292969 Chr.3: 19915317-19915317 on Build GRCh38
rs371282375 Chr.3: 19915348-19915348 on Build GRCh38
rs776421001 Chr.3: 19915376-19915376 on Build GRCh38
rs989686377 Chr.3: 19915377-19915377 on Build GRCh38
rs62279138 Chr.3: 19915503-19915503 on Build GRCh38
rs1055022806 Chr.3: 19915277-19915277 on Build GRCh38
rs376703410 Chr.3: 19915276-19915276 on Build GRCh38
rs760634050 Chr.3: 19915360-19915360 on Build GRCh38
rs9850421 Chr.3: 19915470-19915470 on Build GRCh38
rs761452995 Chr.3: 19915379-19915379 on Build GRCh38
rs373893654 Chr.3: 19915407-19915407 on Build GRCh38
rs751993058 Chr.3: 19915302-19915302 on Build GRCh38
rs539868731 Chr.3: 19915339-19915339 on Build GRCh38

View Full Product Details