Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46653708-46653969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF1
Entrez Gene ID
64756
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
ATP11, ATP11p
UniGene
Hs.100874
Gene Chromosome Location
Chr.1: 46635039-46668427 on Build GRCh38
Transcripts
NM_001042546.2 NM_001243728.1 NM_001256418.1 NM_022745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381141 c.538C>T p.P180S Chr.1: 46653819-46653819 on Build GRCh38
1748484 c.498G>A p.Q166Q Chr.1: 46653859-46653859 on Build GRCh38
1474045 c.525C>T p.V175V Chr.1: 46653832-46653832 on Build GRCh38
4664595 c.511G>A p.A171T Chr.1: 46653846-46653846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771029568 Chr.1: 46653880-46653880 on Build GRCh38
rs753733066 Chr.1: 46653856-46653856 on Build GRCh38
rs532268293 Chr.1: 46653780-46653780 on Build GRCh38
rs941744977 Chr.1: 46653741-46653741 on Build GRCh38
rs773974690 Chr.1: 46653800-46653800 on Build GRCh38
rs766181455 Chr.1: 46653829-46653829 on Build GRCh38
rs761286529 Chr.1: 46653809-46653809 on Build GRCh38
rs1002586088 Chr.1: 46653868-46653868 on Build GRCh38
rs918880949 Chr.1: 46653782-46653782 on Build GRCh38
rs72679900 Chr.1: 46653909-46653909 on Build GRCh38
rs371196515 Chr.1: 46653789-46653789 on Build GRCh38
rs1011935909 Chr.1: 46653768-46653768 on Build GRCh38
rs749066885 Chr.1: 46653873-46653873 on Build GRCh38
rs748259655 Chr.1: 46653883-46653883 on Build GRCh38
rs889061966 Chr.1: 46653874-46653874 on Build GRCh38
rs367951336 Chr.1: 46653828-46653828 on Build GRCh38
rs750045160 Chr.1: 46653825-46653825 on Build GRCh38
rs201480006 Chr.1: 46653857-46653857 on Build GRCh38
rs767194914 Chr.1: 46653812-46653812 on Build GRCh38
rs774385315 Chr.1: 46653902-46653902 on Build GRCh38
rs758417409 Chr.1: 46653899-46653899 on Build GRCh38
rs150528437 Chr.1: 46653931-46653931 on Build GRCh38
rs141637108 Chr.1: 46653878-46653878 on Build GRCh38
rs148411950 Chr.1: 46653859-46653859 on Build GRCh38
rs760153942 Chr.1: 46653783-46653783 on Build GRCh38

View Full Product Details