Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 98858811-98859052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1
Entrez Gene ID
1105
Gene Name
chromodomain helicase DNA binding protein 1
Gene Aliases
-
UniGene
Hs.643465
Gene Chromosome Location
Chr.5: 98855204-98929014 on Build GRCh38
Transcripts
NM_001270.2 XM_005271866.4 XM_005271867.4 XM_011543112.2 XM_017008991.1 XM_017008992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441520 c.4574C>G p.P1525R Chr.5: 98858966-98858966 on Build GRCh38
4169922 c.4562T>C p.V1521A Chr.5: 98858978-98858978 on Build GRCh38
1186796 c.4556C>G p.P1519R Chr.5: 98858984-98858984 on Build GRCh38
5404069 c.4574C>T p.P1525L Chr.5: 98858966-98858966 on Build GRCh38
7106685 c.4525-1G>C p.? Chr.5: 98859016-98859016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751382695 Chr.5: 98858914-98858915 on Build GRCh38
rs767617797 Chr.5: 98858923-98858923 on Build GRCh38
rs765508910 Chr.5: 98858966-98858966 on Build GRCh38
rs1041089174 Chr.5: 98858922-98858922 on Build GRCh38
rs114899267 Chr.5: 98858930-98858930 on Build GRCh38
rs753964035 Chr.5: 98858946-98858946 on Build GRCh38
rs552138105 Chr.5: 98858911-98858911 on Build GRCh38
rs202064808 Chr.5: 98858949-98858949 on Build GRCh38
rs555853100 Chr.5: 98858923-98858923 on Build GRCh38
rs553111541 Chr.5: 98859005-98859005 on Build GRCh38
rs778060142 Chr.5: 98858979-98858979 on Build GRCh38
rs769123490 Chr.5: 98858997-98858997 on Build GRCh38
rs752107996 Chr.5: 98858980-98858980 on Build GRCh38
rs766079748 Chr.5: 98858894-98858897 on Build GRCh38
rs752794119 Chr.5: 98858967-98858967 on Build GRCh38
rs147972187 Chr.5: 98858915-98858915 on Build GRCh38
rs141802026 Chr.5: 98858978-98858978 on Build GRCh38
rs374166699 Chr.5: 98858988-98858988 on Build GRCh38
rs377063711 Chr.5: 98859001-98859001 on Build GRCh38
rs904589216 Chr.5: 98858961-98858961 on Build GRCh38
rs2545734 Chr.5: 98858914-98858914 on Build GRCh38
rs750505760 Chr.5: 98858922-98858923 on Build GRCh38
rs757251660 Chr.5: 98858915-98858915 on Build GRCh38
rs2545735 Chr.5: 98858885-98858885 on Build GRCh38
rs768299632 Chr.5: 98858920-98858920 on Build GRCh38
rs566334849 Chr.5: 98858845-98858845 on Build GRCh38
rs761822213 Chr.5: 98858924-98858924 on Build GRCh38
rs780744455 Chr.5: 98858984-98858984 on Build GRCh38

View Full Product Details