Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49367359-49367624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022063.1 XM_017022064.1

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
UniGene
Hs.122006
Gene Chromosome Location
Chr.15: 49155757-49367883 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521321.2 XM_011521322.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022063.1 XM_017022064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5462466 c.1250T>C p.L417P Chr.15: 49367469-49367469 on Build GRCh38
401551 c.1150C>A p.L384I Chr.15: 49367569-49367569 on Build GRCh38
3501880 c.1137G>A p.E379E Chr.15: 49367582-49367582 on Build GRCh38
3501879 c.1165G>A p.G389S Chr.15: 49367554-49367554 on Build GRCh38
7108142 c.1119T>C p.Y373Y Chr.15: 49367600-49367600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775438426 Chr.15: 49367457-49367457 on Build GRCh38
rs371416601 Chr.15: 49367513-49367513 on Build GRCh38
rs763001854 Chr.15: 49367470-49367470 on Build GRCh38
rs770591797 Chr.15: 49367404-49367404 on Build GRCh38
rs535120047 Chr.15: 49367420-49367420 on Build GRCh38
rs778305075 Chr.15: 49367534-49367534 on Build GRCh38
rs756585144 Chr.15: 49367531-49367531 on Build GRCh38
rs770858827 Chr.15: 49367576-49367576 on Build GRCh38
rs750950812 Chr.15: 49367582-49367582 on Build GRCh38
rs771777388 Chr.15: 49367558-49367558 on Build GRCh38
rs768555380 Chr.15: 49367563-49367563 on Build GRCh38
rs768743764 Chr.15: 49367462-49367462 on Build GRCh38
rs748839118 Chr.15: 49367401-49367401 on Build GRCh38
rs750834633 Chr.15: 49367400-49367415 on Build GRCh38
rs760640496 Chr.15: 49367461-49367461 on Build GRCh38
rs751665404 Chr.15: 49367478-49367478 on Build GRCh38
rs756782281 Chr.15: 49367544-49367544 on Build GRCh38
rs745451388 Chr.15: 49367550-49367550 on Build GRCh38
rs200160678 Chr.15: 49367517-49367517 on Build GRCh38
rs139377249 Chr.15: 49367574-49367574 on Build GRCh38
rs773393896 Chr.15: 49367467-49367467 on Build GRCh38
rs778486123 Chr.15: 49367549-49367549 on Build GRCh38
rs377727825 Chr.15: 49367444-49367444 on Build GRCh38
rs200556221 Chr.15: 49367562-49367562 on Build GRCh38
rs745667919 Chr.15: 49367434-49367434 on Build GRCh38
rs774513018 Chr.15: 49367596-49367596 on Build GRCh38
rs532275129 Chr.15: 49367593-49367594 on Build GRCh38
rs781332253 Chr.15: 49367595-49367595 on Build GRCh38
rs141588026 Chr.15: 49367564-49367564 on Build GRCh38
rs201015191 Chr.15: 49367471-49367471 on Build GRCh38
rs760190246 Chr.15: 49367586-49367586 on Build GRCh38
rs150895583 Chr.15: 49367571-49367571 on Build GRCh38
rs759818247 Chr.15: 49367480-49367480 on Build GRCh38
rs753372861 Chr.15: 49367542-49367542 on Build GRCh38
rs761817125 Chr.15: 49367575-49367575 on Build GRCh38

View Full Product Details