Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6925820-6926057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFA6
Entrez Gene ID
1671
Gene Name
defensin alpha 6
Gene Aliases
DEF6, HD-6
UniGene
Hs.711
Gene Chromosome Location
Chr.8: 6924694-6926076 on Build GRCh38
Transcripts
NM_001926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530352 c.161C>G p.A54G Chr.8: 6925875-6925875 on Build GRCh38
7298364 c.170C>T p.A57V Chr.8: 6925866-6925866 on Build GRCh38
1755774 c.98C>T p.A33V Chr.8: 6925938-6925938 on Build GRCh38
6406097 c.48G>T p.Q16H Chr.8: 6925988-6925988 on Build GRCh38
6414009 c.164A>G p.E55G Chr.8: 6925872-6925872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774663521 Chr.8: 6925924-6925924 on Build GRCh38
rs369140471 Chr.8: 6925916-6925916 on Build GRCh38
rs139059860 Chr.8: 6925885-6925885 on Build GRCh38
rs376197932 Chr.8: 6925911-6925911 on Build GRCh38
rs771978737 Chr.8: 6925929-6925929 on Build GRCh38
rs769999691 Chr.8: 6925886-6925886 on Build GRCh38
rs761941155 Chr.8: 6925896-6925896 on Build GRCh38
rs567531860 Chr.8: 6925867-6925867 on Build GRCh38
rs762700462 Chr.8: 6926011-6926011 on Build GRCh38
rs13439322 Chr.8: 6925937-6925937 on Build GRCh38
rs749833157 Chr.8: 6925961-6925961 on Build GRCh38
rs781513403 Chr.8: 6925881-6925881 on Build GRCh38
rs765041662 Chr.8: 6925939-6925939 on Build GRCh38
rs751078080 Chr.8: 6926002-6926002 on Build GRCh38
rs940107324 Chr.8: 6925980-6925980 on Build GRCh38
rs763767997 Chr.8: 6926015-6926015 on Build GRCh38
rs373665080 Chr.8: 6926024-6926024 on Build GRCh38
rs146649678 Chr.8: 6925915-6925915 on Build GRCh38
rs371286175 Chr.8: 6925905-6925905 on Build GRCh38
rs75695501 Chr.8: 6926031-6926031 on Build GRCh38
rs747081155 Chr.8: 6925996-6925996 on Build GRCh38
rs755815109 Chr.8: 6925862-6925862 on Build GRCh38
rs778595451 Chr.8: 6925873-6925873 on Build GRCh38
rs753617699 Chr.8: 6925908-6925908 on Build GRCh38
rs148947924 Chr.8: 6925949-6925949 on Build GRCh38
rs898459728 Chr.8: 6925855-6925855 on Build GRCh38
rs891362093 Chr.8: 6925876-6925876 on Build GRCh38
rs116451664 Chr.8: 6925898-6925898 on Build GRCh38
rs751198763 Chr.8: 6926016-6926016 on Build GRCh38
rs758078290 Chr.8: 6925945-6925945 on Build GRCh38
rs776504983 Chr.8: 6926000-6926000 on Build GRCh38
rs556620786 Chr.8: 6926017-6926017 on Build GRCh38
rs773091163 Chr.8: 6925946-6925946 on Build GRCh38
rs1049988432 Chr.8: 6925871-6925871 on Build GRCh38
rs45479905 Chr.8: 6925957-6925957 on Build GRCh38
rs768974672 Chr.8: 6925888-6925888 on Build GRCh38
rs774751195 Chr.8: 6925892-6925892 on Build GRCh38
rs755335114 Chr.8: 6925965-6925965 on Build GRCh38
rs555539313 Chr.8: 6925999-6925999 on Build GRCh38
rs762030666 Chr.8: 6925851-6925851 on Build GRCh38
rs753084539 Chr.8: 6926038-6926038 on Build GRCh38
rs151034040 Chr.8: 6926013-6926013 on Build GRCh38
rs750269444 Chr.8: 6925899-6925899 on Build GRCh38
rs767600527 Chr.8: 6925876-6925876 on Build GRCh38
rs779911305 Chr.8: 6925865-6925865 on Build GRCh38
rs748526832 Chr.8: 6925972-6925972 on Build GRCh38
rs979287492 Chr.8: 6926037-6926037 on Build GRCh38
rs750404102 Chr.8: 6925857-6925857 on Build GRCh38
rs752380257 Chr.8: 6925913-6925913 on Build GRCh38
rs773355681 Chr.8: 6925988-6925988 on Build GRCh38
rs764473117 Chr.8: 6925846-6925846 on Build GRCh38
rs573399640 Chr.8: 6925900-6925900 on Build GRCh38
rs751054931 Chr.8: 6925948-6925948 on Build GRCh38
rs969213881 Chr.8: 6925895-6925895 on Build GRCh38
rs752292160 Chr.8: 6925943-6925943 on Build GRCh38
rs941815230 Chr.8: 6925938-6925938 on Build GRCh38
rs763286323 Chr.8: 6925844-6925844 on Build GRCh38
rs775476534 Chr.8: 6925841-6925841 on Build GRCh38
rs113837602 Chr.8: 6925931-6925931 on Build GRCh38
rs1020704493 Chr.8: 6926030-6926030 on Build GRCh38
rs1010201183 Chr.8: 6926021-6926021 on Build GRCh38
rs748394113 Chr.8: 6925926-6925926 on Build GRCh38
rs140184169 Chr.8: 6925919-6925919 on Build GRCh38
rs375500648 Chr.8: 6926022-6926022 on Build GRCh38
rs766318283 Chr.8: 6925933-6925933 on Build GRCh38
rs187327243 Chr.8: 6926005-6926005 on Build GRCh38
rs772324941 Chr.8: 6925983-6925983 on Build GRCh38
rs779423678 Chr.8: 6925966-6925966 on Build GRCh38
rs754818063 Chr.8: 6925909-6925909 on Build GRCh38
rs150712376 Chr.8: 6925850-6925850 on Build GRCh38
rs771612528 Chr.8: 6925880-6925880 on Build GRCh38
rs539202273 Chr.8: 6926018-6926018 on Build GRCh38
rs968827041 Chr.8: 6926033-6926033 on Build GRCh38
rs761771350 Chr.8: 6925930-6925930 on Build GRCh38
rs199502585 Chr.8: 6925868-6925868 on Build GRCh38
rs775931969 Chr.8: 6925887-6925887 on Build GRCh38
rs199812447 Chr.8: 6926035-6926035 on Build GRCh38
rs780471666 Chr.8: 6925918-6925918 on Build GRCh38
rs949033323 Chr.8: 6925934-6925934 on Build GRCh38
rs760790034 Chr.8: 6925854-6925854 on Build GRCh38
rs769171181 Chr.8: 6925922-6925922 on Build GRCh38
rs919938266 Chr.8: 6926003-6926003 on Build GRCh38
rs997518344 Chr.8: 6925866-6925866 on Build GRCh38

View Full Product Details