Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179954868-179955121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC22
Entrez Gene ID
57703
Gene Name
CWC22 homolog, spliceosome-associated protein
Gene Aliases
EIF4GL, NCM, fSAPb
UniGene
Hs.311363
Gene Chromosome Location
Chr.2: 179944876-180007359 on Build GRCh38
Transcripts
NM_020943.2 XM_005246726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242166 c.1515A>T p.K505N Chr.2: 179954978-179954978 on Build GRCh38
5353892 c.1459-10delT p.? Chr.2: 179955044-179955044 on Build GRCh38
7262449 c.1459-10_1459-9insT p.? Chr.2: 179955043-179955044 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371835416 Chr.2: 179954912-179954912 on Build GRCh38
rs948341582 Chr.2: 179954920-179954920 on Build GRCh38
rs776187974 Chr.2: 179954940-179954940 on Build GRCh38
rs756473219 Chr.2: 179954984-179954984 on Build GRCh38
rs373590794 Chr.2: 179955078-179955078 on Build GRCh38
rs768138839 Chr.2: 179954931-179954931 on Build GRCh38
rs767939410 Chr.2: 179954975-179954975 on Build GRCh38
rs756040125 Chr.2: 179955044-179955044 on Build GRCh38
rs752130054 Chr.2: 179954960-179954960 on Build GRCh38
rs757382783 Chr.2: 179955001-179955001 on Build GRCh38
rs200509255 Chr.2: 179955044-179955044 on Build GRCh38
rs543228207 Chr.2: 179955043-179955044 on Build GRCh38
rs915537144 Chr.2: 179954902-179954902 on Build GRCh38
rs1047215004 Chr.2: 179955089-179955089 on Build GRCh38
rs769376647 Chr.2: 179955083-179955083 on Build GRCh38
rs374720550 Chr.2: 179954918-179954918 on Build GRCh38
rs369488955 Chr.2: 179955063-179955063 on Build GRCh38
rs372078508 Chr.2: 179955042-179955042 on Build GRCh38
rs543488270 Chr.2: 179955095-179955095 on Build GRCh38
rs533353756 Chr.2: 179955087-179955087 on Build GRCh38
rs994338172 Chr.2: 179955052-179955052 on Build GRCh38
rs925459151 Chr.2: 179955028-179955028 on Build GRCh38
rs754023853 Chr.2: 179954989-179954989 on Build GRCh38
rs760330542 Chr.2: 179954927-179954927 on Build GRCh38
rs752954929 Chr.2: 179954983-179954983 on Build GRCh38
rs763417774 Chr.2: 179954950-179954950 on Build GRCh38
rs745825620 Chr.2: 179955025-179955025 on Build GRCh38
rs759778072 Chr.2: 179954967-179954967 on Build GRCh38
rs905747209 Chr.2: 179955092-179955092 on Build GRCh38
rs754620333 Chr.2: 179954905-179954905 on Build GRCh38
rs778679787 Chr.2: 179955022-179955022 on Build GRCh38
rs766842052 Chr.2: 179954951-179954951 on Build GRCh38
rs1043858571 Chr.2: 179955027-179955027 on Build GRCh38
rs768334977 Chr.2: 179955053-179955053 on Build GRCh38
rs746657885 Chr.2: 179955050-179955050 on Build GRCh38
rs1013872178 Chr.2: 179955059-179955059 on Build GRCh38
rs368961005 Chr.2: 179954987-179954987 on Build GRCh38

View Full Product Details