Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30272514-30272785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLMH
Entrez Gene ID
642
Gene Name
bleomycin hydrolase
Gene Aliases
BH, BMH
UniGene
Hs.371914
Gene Chromosome Location
Chr.17: 30248195-30292166 on Build GRCh38
Transcripts
NM_000386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295367 c.953A>T p.D318V Chr.17: 30272748-30272748 on Build GRCh38
4513643 c.945C>T p.S315S Chr.17: 30272756-30272756 on Build GRCh38
299820 c.1011C>T p.G337G Chr.17: 30272578-30272578 on Build GRCh38
4450932 c.960+2T>C p.? Chr.17: 30272739-30272739 on Build GRCh38
353939 c.961G>T p.A321S Chr.17: 30272628-30272628 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746289690 Chr.17: 30272626-30272626 on Build GRCh38
rs576860161 Chr.17: 30272633-30272633 on Build GRCh38
rs200502806 Chr.17: 30272644-30272644 on Build GRCh38
rs775380825 Chr.17: 30272713-30272713 on Build GRCh38
rs184963100 Chr.17: 30272570-30272570 on Build GRCh38
rs764438981 Chr.17: 30272718-30272718 on Build GRCh38
rs759656087 Chr.17: 30272678-30272678 on Build GRCh38
rs75301481 Chr.17: 30272609-30272609 on Build GRCh38
rs368832391 Chr.17: 30272731-30272731 on Build GRCh38
rs748141183 Chr.17: 30272615-30272615 on Build GRCh38
rs201046057 Chr.17: 30272558-30272558 on Build GRCh38
rs769891300 Chr.17: 30272709-30272709 on Build GRCh38
rs755792054 Chr.17: 30272575-30272575 on Build GRCh38
rs766445796 Chr.17: 30272539-30272539 on Build GRCh38
rs372310785 Chr.17: 30272760-30272760 on Build GRCh38
rs935820536 Chr.17: 30272714-30272714 on Build GRCh38
rs763325386 Chr.17: 30272568-30272568 on Build GRCh38
rs776762780 Chr.17: 30272544-30272544 on Build GRCh38
rs771205635 Chr.17: 30272662-30272662 on Build GRCh38
rs375335286 Chr.17: 30272563-30272563 on Build GRCh38
rs752027911 Chr.17: 30272727-30272727 on Build GRCh38
rs765784825 Chr.17: 30272694-30272721 on Build GRCh38
rs754629259 Chr.17: 30272749-30272749 on Build GRCh38
rs368088469 Chr.17: 30272591-30272591 on Build GRCh38
rs751808439 Chr.17: 30272574-30272574 on Build GRCh38
rs776814099 Chr.17: 30272676-30272676 on Build GRCh38
rs779032343 Chr.17: 30272608-30272608 on Build GRCh38
rs759603052 Chr.17: 30272553-30272553 on Build GRCh38
rs766182542 Chr.17: 30272734-30272734 on Build GRCh38
rs891581795 Chr.17: 30272737-30272737 on Build GRCh38
rs116778162 Chr.17: 30272717-30272717 on Build GRCh38
rs147322544 Chr.17: 30272590-30272590 on Build GRCh38
rs889636026 Chr.17: 30272680-30272680 on Build GRCh38
rs746779129 Chr.17: 30272661-30272661 on Build GRCh38
rs753508547 Chr.17: 30272747-30272747 on Build GRCh38
rs762417371 Chr.17: 30272639-30272642 on Build GRCh38
rs754513319 Chr.17: 30272595-30272595 on Build GRCh38

View Full Product Details