Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30968963-30969218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3637961 c.176G>A p.W59* Chr.7: 30969078-30969078 on Build GRCh38
5698551 c.236C>A p.P79Q Chr.7: 30969138-30969138 on Build GRCh38
3637962 c.228C>T p.L76L Chr.7: 30969130-30969130 on Build GRCh38
6566822 c.161-1G>C p.? Chr.7: 30969062-30969062 on Build GRCh38
4984977 c.169G>A p.A57T Chr.7: 30969071-30969071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865904751 Chr.7: 30969170-30969170 on Build GRCh38
rs751885823 Chr.7: 30969166-30969166 on Build GRCh38
rs762629722 Chr.7: 30969043-30969043 on Build GRCh38
rs752793580 Chr.7: 30969072-30969072 on Build GRCh38
rs192109771 Chr.7: 30968991-30968991 on Build GRCh38
rs768112214 Chr.7: 30969115-30969115 on Build GRCh38
rs989948340 Chr.7: 30969151-30969151 on Build GRCh38
rs763709666 Chr.7: 30969051-30969051 on Build GRCh38
rs758585465 Chr.7: 30969150-30969150 on Build GRCh38
rs781077252 Chr.7: 30969177-30969177 on Build GRCh38
rs774857155 Chr.7: 30969041-30969041 on Build GRCh38
rs760129030 Chr.7: 30969135-30969135 on Build GRCh38
rs761056409 Chr.7: 30969054-30969054 on Build GRCh38
rs769403061 Chr.7: 30969019-30969019 on Build GRCh38
rs776859854 Chr.7: 30969132-30969132 on Build GRCh38
rs755430803 Chr.7: 30969067-30969067 on Build GRCh38
rs764209354 Chr.7: 30969155-30969155 on Build GRCh38
rs577536745 Chr.7: 30969099-30969099 on Build GRCh38
rs942863057 Chr.7: 30969002-30969002 on Build GRCh38
rs376235715 Chr.7: 30969014-30969014 on Build GRCh38
rs948242768 Chr.7: 30968997-30968997 on Build GRCh38
rs28371558 Chr.7: 30969139-30969139 on Build GRCh38
rs563078420 Chr.7: 30969103-30969103 on Build GRCh38
rs540740642 Chr.7: 30969076-30969076 on Build GRCh38
rs4988496 Chr.7: 30969071-30969071 on Build GRCh38
rs745989289 Chr.7: 30969001-30969001 on Build GRCh38
rs867537571 Chr.7: 30969097-30969097 on Build GRCh38
rs773941149 Chr.7: 30969053-30969053 on Build GRCh38
rs121918117 Chr.7: 30969116-30969116 on Build GRCh38
rs765640577 Chr.7: 30969138-30969138 on Build GRCh38
rs4988495 Chr.7: 30968992-30968992 on Build GRCh38
rs766587678 Chr.7: 30969063-30969063 on Build GRCh38
rs565445448 Chr.7: 30969091-30969091 on Build GRCh38
rs373112876 Chr.7: 30969011-30969011 on Build GRCh38
rs754386566 Chr.7: 30969066-30969066 on Build GRCh38
rs757412947 Chr.7: 30969171-30969171 on Build GRCh38
rs201893637 Chr.7: 30969102-30969102 on Build GRCh38
rs958079133 Chr.7: 30969148-30969148 on Build GRCh38
rs761399375 Chr.7: 30969121-30969121 on Build GRCh38
rs967674479 Chr.7: 30969191-30969191 on Build GRCh38
rs769587851 Chr.7: 30969109-30969109 on Build GRCh38
rs775025721 Chr.7: 30969110-30969110 on Build GRCh38
rs940355976 Chr.7: 30969036-30969036 on Build GRCh38
rs141243007 Chr.7: 30969194-30969194 on Build GRCh38
rs547825255 Chr.7: 30969128-30969128 on Build GRCh38
rs1050194454 Chr.7: 30969083-30969083 on Build GRCh38
rs369751652 Chr.7: 30969073-30969073 on Build GRCh38
rs745733075 Chr.7: 30969018-30969018 on Build GRCh38
rs745628562 Chr.7: 30969192-30969192 on Build GRCh38
rs994960254 Chr.7: 30969075-30969075 on Build GRCh38

View Full Product Details