Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 69599675-69599924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
UniGene
Hs.21631
Gene Chromosome Location
Chr.7: 69598475-70793068 on Build GRCh38
Transcripts
NM_001127231.2 NM_001127232.2 NM_015570.3 XM_011516010.2 XM_011516011.2 XM_011516012.2 XM_011516013.2 XM_011516014.2 XM_017011951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4646292 c.113G>A p.G38D Chr.7: 69599766-69599766 on Build GRCh38
6608927 c.166A>G p.K56E Chr.7: 69599819-69599819 on Build GRCh38
3086272 c.245C>A p.S82* Chr.7: 69599898-69599898 on Build GRCh38
3086271 c.245C>A p.S82* Chr.7: 69599898-69599898 on Build GRCh38
7284193 c.49C>T p.R17W Chr.7: 69599702-69599702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762676699 Chr.7: 69599755-69599778 on Build GRCh38
rs945733678 Chr.7: 69599781-69599781 on Build GRCh38
rs911513762 Chr.7: 69599771-69599771 on Build GRCh38
rs753526278 Chr.7: 69599863-69599863 on Build GRCh38
rs777541048 Chr.7: 69599782-69599782 on Build GRCh38
rs1041469078 Chr.7: 69599787-69599787 on Build GRCh38
rs752490672 Chr.7: 69599750-69599773 on Build GRCh38
rs756779190 Chr.7: 69599784-69599784 on Build GRCh38
rs746774405 Chr.7: 69599806-69599806 on Build GRCh38
rs1001747687 Chr.7: 69599854-69599854 on Build GRCh38
rs750105505 Chr.7: 69599838-69599838 on Build GRCh38
rs1024257295 Chr.7: 69599858-69599858 on Build GRCh38
rs1004195592 Chr.7: 69599894-69599894 on Build GRCh38
rs756725295 Chr.7: 69599881-69599881 on Build GRCh38
rs780512031 Chr.7: 69599755-69599755 on Build GRCh38
rs775225727 Chr.7: 69599802-69599802 on Build GRCh38
rs761862785 Chr.7: 69599833-69599833 on Build GRCh38
rs755373797 Chr.7: 69599845-69599845 on Build GRCh38
rs776816196 Chr.7: 69599820-69599820 on Build GRCh38
rs910011165 Chr.7: 69599731-69599731 on Build GRCh38
rs771784577 Chr.7: 69599801-69599801 on Build GRCh38
rs763898324 Chr.7: 69599853-69599853 on Build GRCh38
rs552587555 Chr.7: 69599836-69599836 on Build GRCh38
rs1051354561 Chr.7: 69599761-69599761 on Build GRCh38
rs976017092 Chr.7: 69599743-69599743 on Build GRCh38
rs745753819 Chr.7: 69599798-69599798 on Build GRCh38
rs752423410 Chr.7: 69599765-69599765 on Build GRCh38
rs755792920 Chr.7: 69599780-69599780 on Build GRCh38
rs913230763 Chr.7: 69599727-69599727 on Build GRCh38
rs934214411 Chr.7: 69599759-69599759 on Build GRCh38
rs530867061 Chr.7: 69599740-69599740 on Build GRCh38
rs748798665 Chr.7: 69599783-69599783 on Build GRCh38
rs946041123 Chr.7: 69599815-69599815 on Build GRCh38
rs962834047 Chr.7: 69599733-69599733 on Build GRCh38
rs768911898 Chr.7: 69599810-69599810 on Build GRCh38
rs199945820 Chr.7: 69599841-69599841 on Build GRCh38
rs904301389 Chr.7: 69599794-69599794 on Build GRCh38
rs778785768 Chr.7: 69599797-69599797 on Build GRCh38

View Full Product Details