Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83057207-83057480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS4
Entrez Gene ID
51138
Gene Name
COP9 signalosome subunit 4
Gene Aliases
CSN4
UniGene
Hs.190384
Gene Chromosome Location
Chr.4: 83035086-83075818 on Build GRCh38
Transcripts
NM_001258006.1 NM_016129.2 XM_011532019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634091 c.697A>G p.T233A Chr.4: 83057390-83057390 on Build GRCh38
6634090 c.714A>G p.A238A Chr.4: 83057407-83057407 on Build GRCh38
345661 c.586G>C p.D196H Chr.4: 83057279-83057279 on Build GRCh38
4758279 c.686C>G p.A229G Chr.4: 83057379-83057379 on Build GRCh38
6545572 c.699G>T p.T233T Chr.4: 83057392-83057392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781036672 Chr.4: 83057314-83057314 on Build GRCh38
rs749216255 Chr.4: 83057418-83057418 on Build GRCh38
rs375741322 Chr.4: 83057260-83057260 on Build GRCh38
rs745710271 Chr.4: 83057318-83057318 on Build GRCh38
rs771622016 Chr.4: 83057436-83057436 on Build GRCh38
rs777856541 Chr.4: 83057299-83057299 on Build GRCh38
rs779357087 Chr.4: 83057326-83057326 on Build GRCh38
rs373322675 Chr.4: 83057253-83057253 on Build GRCh38
rs535518390 Chr.4: 83057405-83057405 on Build GRCh38
rs774049558 Chr.4: 83057339-83057339 on Build GRCh38
rs747768262 Chr.4: 83057343-83057343 on Build GRCh38
rs370098037 Chr.4: 83057377-83057377 on Build GRCh38
rs748934323 Chr.4: 83057380-83057380 on Build GRCh38
rs906864158 Chr.4: 83057446-83057446 on Build GRCh38
rs796066640 Chr.4: 83057304-83057304 on Build GRCh38
rs747715219 Chr.4: 83057430-83057430 on Build GRCh38
rs772703468 Chr.4: 83057442-83057442 on Build GRCh38
rs750660159 Chr.4: 83057415-83057415 on Build GRCh38
rs377401913 Chr.4: 83057417-83057417 on Build GRCh38
rs751762432 Chr.4: 83057308-83057308 on Build GRCh38
rs765668530 Chr.4: 83057366-83057366 on Build GRCh38
rs373129606 Chr.4: 83057414-83057414 on Build GRCh38
rs755002006 Chr.4: 83057419-83057419 on Build GRCh38
rs371262270 Chr.4: 83057435-83057435 on Build GRCh38
rs775815859 Chr.4: 83057372-83057372 on Build GRCh38
rs753254618 Chr.4: 83057270-83057270 on Build GRCh38
rs200687602 Chr.4: 83057251-83057251 on Build GRCh38
rs111735566 Chr.4: 83057427-83057427 on Build GRCh38
rs768231411 Chr.4: 83057334-83057334 on Build GRCh38
rs759755279 Chr.4: 83057362-83057362 on Build GRCh38
rs777038755 Chr.4: 83057352-83057352 on Build GRCh38
rs777264949 Chr.4: 83057278-83057278 on Build GRCh38
rs771914503 Chr.4: 83057347-83057347 on Build GRCh38
rs757511849 Chr.4: 83057311-83057311 on Build GRCh38
rs755952178 Chr.4: 83057319-83057319 on Build GRCh38
rs574807433 Chr.4: 83057416-83057416 on Build GRCh38
rs763503741 Chr.4: 83057374-83057374 on Build GRCh38
rs948447458 Chr.4: 83057434-83057434 on Build GRCh38

View Full Product Details