Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45027586-45027857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2 XM_017012671.1 XM_017012672.1 XM_017012673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3263222 c.17G>A p.R6Q Chr.7: 45027721-45027721 on Build GRCh38
1755298 c.45A>C p.K15N Chr.7: 45027749-45027749 on Build GRCh38
6561424 c.52C>A p.P18T Chr.7: 45027756-45027756 on Build GRCh38
1755299 c.55C>T p.Q19* Chr.7: 45027759-45027759 on Build GRCh38
5799078 c.30C>G p.N10K Chr.7: 45027734-45027734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373357366 Chr.7: 45027658-45027658 on Build GRCh38
rs761216215 Chr.7: 45027753-45027753 on Build GRCh38
rs772761867 Chr.7: 45027766-45027766 on Build GRCh38
rs550045282 Chr.7: 45027780-45027780 on Build GRCh38
rs752527164 Chr.7: 45027790-45027790 on Build GRCh38
rs762310131 Chr.7: 45027772-45027772 on Build GRCh38
rs746124946 Chr.7: 45027736-45027736 on Build GRCh38
rs775996355 Chr.7: 45027823-45027823 on Build GRCh38
rs371886141 Chr.7: 45027683-45027683 on Build GRCh38
rs756066541 Chr.7: 45027797-45027797 on Build GRCh38
rs376057553 Chr.7: 45027708-45027708 on Build GRCh38
rs779160853 Chr.7: 45027814-45027814 on Build GRCh38
rs1014245661 Chr.7: 45027804-45027804 on Build GRCh38
rs779628972 Chr.7: 45027682-45027682 on Build GRCh38
rs749387943 Chr.7: 45027721-45027721 on Build GRCh38
rs923458913 Chr.7: 45027640-45027640 on Build GRCh38
rs777515756 Chr.7: 45027801-45027801 on Build GRCh38
rs960023676 Chr.7: 45027809-45027809 on Build GRCh38
rs768000149 Chr.7: 45027654-45027654 on Build GRCh38
rs755722007 Chr.7: 45027813-45027813 on Build GRCh38
rs147668138 Chr.7: 45027715-45027715 on Build GRCh38
rs2289365 Chr.7: 45027668-45027668 on Build GRCh38
rs769076048 Chr.7: 45027834-45027834 on Build GRCh38
rs527794718 Chr.7: 45027646-45027646 on Build GRCh38
rs769667526 Chr.7: 45027718-45027718 on Build GRCh38
rs1002714055 Chr.7: 45027833-45027833 on Build GRCh38
rs560338124 Chr.7: 45027609-45027609 on Build GRCh38
rs904462838 Chr.7: 45027740-45027740 on Build GRCh38
rs772520097 Chr.7: 45027739-45027739 on Build GRCh38
rs372486302 Chr.7: 45027787-45027787 on Build GRCh38
rs376876775 Chr.7: 45027720-45027720 on Build GRCh38
rs747388126 Chr.7: 45027824-45027824 on Build GRCh38
rs746069445 Chr.7: 45027817-45027817 on Build GRCh38
rs772465603 Chr.7: 45027822-45027822 on Build GRCh38
rs761290961 Chr.7: 45027666-45027666 on Build GRCh38
rs776049858 Chr.7: 45027748-45027748 on Build GRCh38
rs749253736 Chr.7: 45027802-45027802 on Build GRCh38
rs765914345 Chr.7: 45027776-45027776 on Build GRCh38
rs942032703 Chr.7: 45027697-45027697 on Build GRCh38
rs886527726 Chr.7: 45027690-45027690 on Build GRCh38
rs1012808303 Chr.7: 45027733-45027733 on Build GRCh38
rs369487153 Chr.7: 45027730-45027730 on Build GRCh38
rs767060958 Chr.7: 45027608-45027608 on Build GRCh38
rs754697580 Chr.7: 45027709-45027709 on Build GRCh38
rs891637905 Chr.7: 45027711-45027711 on Build GRCh38
rs1048247360 Chr.7: 45027684-45027684 on Build GRCh38
rs1001582686 Chr.7: 45027761-45027761 on Build GRCh38
rs764578298 Chr.7: 45027763-45027763 on Build GRCh38
rs780995903 Chr.7: 45027713-45027713 on Build GRCh38
rs758062005 Chr.7: 45027669-45027669 on Build GRCh38
rs77090093 Chr.7: 45027732-45027732 on Build GRCh38
rs754646521 Chr.7: 45027786-45027786 on Build GRCh38

View Full Product Details