Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154284636-154284871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX36
Entrez Gene ID
170506
Gene Name
DEAH-box helicase 36
Gene Aliases
DDX36, G4R1, MLEL1, RHAU
UniGene
Hs.446270
Gene Chromosome Location
Chr.3: 154275668-154324497 on Build GRCh38
Transcripts
NM_001114397.1 NM_020865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4114619 c.2174A>C p.K725T Chr.3: 154284659-154284659 on Build GRCh38
4773634 c.2163+7A>G p.? Chr.3: 154284807-154284807 on Build GRCh38
4114618 c.2216A>C p.K739T Chr.3: 154284659-154284659 on Build GRCh38
479604 c.2235A>G p.R745R Chr.3: 154284640-154284640 on Build GRCh38
7259728 c.2171A>T p.E724V Chr.3: 154284662-154284662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750327747 Chr.3: 154284717-154284717 on Build GRCh38
rs765145539 Chr.3: 154284709-154284709 on Build GRCh38
rs749716227 Chr.3: 154284811-154284811 on Build GRCh38
rs756425679 Chr.3: 154284795-154284795 on Build GRCh38
rs1050556288 Chr.3: 154284746-154284746 on Build GRCh38
rs1043187850 Chr.3: 154284822-154284822 on Build GRCh38
rs751069370 Chr.3: 154284783-154284783 on Build GRCh38
rs897788702 Chr.3: 154284688-154284688 on Build GRCh38
rs1006752629 Chr.3: 154284662-154284662 on Build GRCh38
rs770152596 Chr.3: 154284693-154284694 on Build GRCh38
rs1025202627 Chr.3: 154284724-154284724 on Build GRCh38
rs140951576 Chr.3: 154284814-154284814 on Build GRCh38
rs777946334 Chr.3: 154284727-154284727 on Build GRCh38
rs374691038 Chr.3: 154284829-154284829 on Build GRCh38
rs370305174 Chr.3: 154284825-154284825 on Build GRCh38
rs923560979 Chr.3: 154284714-154284714 on Build GRCh38
rs751743467 Chr.3: 154284793-154284793 on Build GRCh38
rs754497468 Chr.3: 154284787-154284787 on Build GRCh38
rs373836205 Chr.3: 154284790-154284790 on Build GRCh38
rs567380128 Chr.3: 154284721-154284721 on Build GRCh38
rs775729954 Chr.3: 154284848-154284848 on Build GRCh38
rs758415035 Chr.3: 154284718-154284718 on Build GRCh38
rs768176187 Chr.3: 154284836-154284836 on Build GRCh38
rs199954319 Chr.3: 154284846-154284846 on Build GRCh38
rs200507247 Chr.3: 154284687-154284687 on Build GRCh38
rs188695285 Chr.3: 154284749-154284749 on Build GRCh38
rs761627141 Chr.3: 154284703-154284703 on Build GRCh38
rs62276969 Chr.3: 154284808-154284808 on Build GRCh38

View Full Product Details