Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31414748-31415011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
UniGene
Hs.187269
Gene Chromosome Location
Chr.11: 31369829-31432835 on Build GRCh38
Transcripts
NM_181706.4 XM_017017216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4678748 c.245G>A p.R82Q Chr.11: 31414944-31414944 on Build GRCh38
1250453 c.240G>A p.L80L Chr.11: 31414939-31414939 on Build GRCh38
122655 c.250+3A>G p.? Chr.11: 31414952-31414952 on Build GRCh38
6045585 c.137A>T p.D46V Chr.11: 31414836-31414836 on Build GRCh38
4969426 c.222_223insC p.K75fs*5 Chr.11: 31414921-31414922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779845550 Chr.11: 31414944-31414944 on Build GRCh38
rs559918901 Chr.11: 31414943-31414943 on Build GRCh38
rs369721738 Chr.11: 31414825-31414825 on Build GRCh38
rs756061639 Chr.11: 31414776-31414776 on Build GRCh38
rs201394853 Chr.11: 31414982-31414982 on Build GRCh38
rs771209942 Chr.11: 31414974-31414974 on Build GRCh38
rs1036572038 Chr.11: 31414886-31414886 on Build GRCh38
rs373436499 Chr.11: 31414827-31414827 on Build GRCh38
rs772113160 Chr.11: 31414983-31414983 on Build GRCh38
rs748632715 Chr.11: 31414954-31414954 on Build GRCh38
rs375514504 Chr.11: 31414892-31414892 on Build GRCh38
rs770285026 Chr.11: 31414956-31414956 on Build GRCh38
rs947718550 Chr.11: 31414777-31414777 on Build GRCh38
rs777002044 Chr.11: 31414823-31414826 on Build GRCh38
rs766823469 Chr.11: 31414794-31414799 on Build GRCh38
rs757119357 Chr.11: 31414922-31414922 on Build GRCh38
rs201981981 Chr.11: 31414910-31414910 on Build GRCh38
rs1048727886 Chr.11: 31414788-31414788 on Build GRCh38
rs368900665 Chr.11: 31414913-31414913 on Build GRCh38
rs750187354 Chr.11: 31414938-31414938 on Build GRCh38
rs775601185 Chr.11: 31414984-31414984 on Build GRCh38
rs749589524 Chr.11: 31414969-31414969 on Build GRCh38
rs554061713 Chr.11: 31414839-31414839 on Build GRCh38
rs372428770 Chr.11: 31414932-31414932 on Build GRCh38
rs538538453 Chr.11: 31414820-31414820 on Build GRCh38
rs11552244 Chr.11: 31414877-31414877 on Build GRCh38
rs756992866 Chr.11: 31414808-31414808 on Build GRCh38
rs763952784 Chr.11: 31414785-31414785 on Build GRCh38
rs753734622 Chr.11: 31414801-31414801 on Build GRCh38
rs778013457 Chr.11: 31414960-31414960 on Build GRCh38
rs770022608 Chr.11: 31414863-31414863 on Build GRCh38
rs751168624 Chr.11: 31414945-31414945 on Build GRCh38
rs753700947 Chr.11: 31414918-31414918 on Build GRCh38
rs376175939 Chr.11: 31414950-31414950 on Build GRCh38
rs761681016 Chr.11: 31414987-31414987 on Build GRCh38
rs748594924 Chr.11: 31414854-31414854 on Build GRCh38
rs776986887 Chr.11: 31414840-31414840 on Build GRCh38
rs146731433 Chr.11: 31414988-31414988 on Build GRCh38
rs747408168 Chr.11: 31414838-31414838 on Build GRCh38
rs764077492 Chr.11: 31414914-31414914 on Build GRCh38
rs144832808 Chr.11: 31414980-31414980 on Build GRCh38
rs763105799 Chr.11: 31414895-31414895 on Build GRCh38
rs746450440 Chr.11: 31414835-31414835 on Build GRCh38
rs774543304 Chr.11: 31414978-31414978 on Build GRCh38
rs766531883 Chr.11: 31414897-31414897 on Build GRCh38
rs778680263 Chr.11: 31414812-31414812 on Build GRCh38
rs759862339 Chr.11: 31414925-31414926 on Build GRCh38

View Full Product Details