Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38234465-38234690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
UniGene
Hs.434966
Gene Chromosome Location
Chr.8: 38231343-38273442 on Build GRCh38
Transcripts
NM_001164232.1 NM_001164234.1 NM_015214.2 XM_005273454.2 XM_005273455.3 XM_005273456.3 XM_011544456.1 XM_017013252.1 XM_017013253.1 XM_017013254.1 XM_017013255.1 XM_017013256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6472574 c.385T>C p.S129P Chr.8: 38234558-38234558 on Build GRCh38
376925 c.322T>G p.S108A Chr.8: 38234495-38234495 on Build GRCh38
5358783 c.342G>A p.T114T Chr.8: 38234515-38234515 on Build GRCh38
6254693 c.310G>T p.D104Y Chr.8: 38234483-38234483 on Build GRCh38
1699986 c.379C>T p.P127S Chr.8: 38234552-38234552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777773204 Chr.8: 38234587-38234587 on Build GRCh38
rs564894062 Chr.8: 38234633-38234633 on Build GRCh38
rs775275421 Chr.8: 38234532-38234532 on Build GRCh38
rs141974093 Chr.8: 38234585-38234585 on Build GRCh38
rs755352037 Chr.8: 38234502-38234502 on Build GRCh38
rs746829853 Chr.8: 38234607-38234607 on Build GRCh38
rs144993452 Chr.8: 38234541-38234541 on Build GRCh38
rs748280169 Chr.8: 38234511-38234511 on Build GRCh38
rs562827417 Chr.8: 38234590-38234590 on Build GRCh38
rs757206544 Chr.8: 38234612-38234615 on Build GRCh38
rs748398783 Chr.8: 38234559-38234559 on Build GRCh38
rs774309381 Chr.8: 38234515-38234515 on Build GRCh38
rs201640343 Chr.8: 38234548-38234548 on Build GRCh38
rs973316510 Chr.8: 38234618-38234618 on Build GRCh38
rs750939885 Chr.8: 38234552-38234552 on Build GRCh38
rs766794813 Chr.8: 38234560-38234560 on Build GRCh38
rs377264637 Chr.8: 38234611-38234611 on Build GRCh38
rs780636754 Chr.8: 38234497-38234497 on Build GRCh38
rs771841303 Chr.8: 38234530-38234530 on Build GRCh38
rs201121892 Chr.8: 38234566-38234566 on Build GRCh38
rs148587720 Chr.8: 38234533-38234533 on Build GRCh38
rs767446112 Chr.8: 38234625-38234628 on Build GRCh38
rs751458468 Chr.8: 38234607-38234608 on Build GRCh38
rs191787470 Chr.8: 38234543-38234543 on Build GRCh38
rs768385527 Chr.8: 38234613-38234613 on Build GRCh38
rs920531203 Chr.8: 38234662-38234662 on Build GRCh38
rs745871510 Chr.8: 38234517-38234517 on Build GRCh38
rs572255417 Chr.8: 38234496-38234496 on Build GRCh38
rs779785705 Chr.8: 38234602-38234602 on Build GRCh38
rs750941346 Chr.8: 38234494-38234494 on Build GRCh38
rs781089161 Chr.8: 38234508-38234508 on Build GRCh38
rs776316461 Chr.8: 38234620-38234620 on Build GRCh38
rs542604783 Chr.8: 38234514-38234514 on Build GRCh38
rs755200059 Chr.8: 38234567-38234567 on Build GRCh38
rs745746575 Chr.8: 38234589-38234589 on Build GRCh38
rs755267771 Chr.8: 38234507-38234507 on Build GRCh38
rs777774654 Chr.8: 38234574-38234574 on Build GRCh38
rs1016775180 Chr.8: 38234660-38234660 on Build GRCh38
rs764694270 Chr.8: 38234550-38234550 on Build GRCh38
rs772199158 Chr.8: 38234573-38234573 on Build GRCh38

View Full Product Details