Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35749932-35750193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L2
Entrez Gene ID
246176
Gene Name
growth arrest specific 2 like 2
Gene Aliases
GAR17
UniGene
Hs.591194
Gene Chromosome Location
Chr.17: 35744511-35752878 on Build GRCh38
Transcripts
NM_139285.3

SNP Detail

SNP ID Chromosome Location
rs782429430 Chr.17: 35750085-35750085 on Build GRCh38
rs782805661 Chr.17: 35750087-35750087 on Build GRCh38
rs965224107 Chr.17: 35749964-35749964 on Build GRCh38
rs782698841 Chr.17: 35750124-35750124 on Build GRCh38
rs950242380 Chr.17: 35750136-35750136 on Build GRCh38
rs782576743 Chr.17: 35750106-35750106 on Build GRCh38
rs782489530 Chr.17: 35750104-35750104 on Build GRCh38
rs978424560 Chr.17: 35750033-35750033 on Build GRCh38
rs781981127 Chr.17: 35750053-35750053 on Build GRCh38
rs368613685 Chr.17: 35750122-35750122 on Build GRCh38
rs587609118 Chr.17: 35750139-35750139 on Build GRCh38
rs782721203 Chr.17: 35750077-35750077 on Build GRCh38
rs782649031 Chr.17: 35750101-35750101 on Build GRCh38
rs781921684 Chr.17: 35750044-35750044 on Build GRCh38
rs782612691 Chr.17: 35750167-35750167 on Build GRCh38
rs782230260 Chr.17: 35750103-35750103 on Build GRCh38
rs587640921 Chr.17: 35750043-35750044 on Build GRCh38
rs781811296 Chr.17: 35750147-35750147 on Build GRCh38
rs71381471 Chr.17: 35750173-35750173 on Build GRCh38
rs782244176 Chr.17: 35750116-35750116 on Build GRCh38
rs150421836 Chr.17: 35750145-35750145 on Build GRCh38
rs781953251 Chr.17: 35750121-35750121 on Build GRCh38
rs782329384 Chr.17: 35750041-35750041 on Build GRCh38
rs368099080 Chr.17: 35750047-35750047 on Build GRCh38
rs781892521 Chr.17: 35750160-35750160 on Build GRCh38
rs782023779 Chr.17: 35750111-35750111 on Build GRCh38
rs782352046 Chr.17: 35750118-35750118 on Build GRCh38
rs781864159 Chr.17: 35750140-35750140 on Build GRCh38
rs782062557 Chr.17: 35750123-35750123 on Build GRCh38
rs376528605 Chr.17: 35750093-35750093 on Build GRCh38
rs782799202 Chr.17: 35750052-35750052 on Build GRCh38
rs782145751 Chr.17: 35750131-35750131 on Build GRCh38
rs782525112 Chr.17: 35750165-35750165 on Build GRCh38
rs782524428 Chr.17: 35750094-35750094 on Build GRCh38
rs1020026089 Chr.17: 35749984-35749984 on Build GRCh38
rs782106683 Chr.17: 35750143-35750143 on Build GRCh38
rs372298233 Chr.17: 35750129-35750129 on Build GRCh38
rs958956381 Chr.17: 35750120-35750120 on Build GRCh38
rs782095789 Chr.17: 35750069-35750069 on Build GRCh38
rs138166711 Chr.17: 35750152-35750152 on Build GRCh38
rs917428094 Chr.17: 35750128-35750128 on Build GRCh38
rs967500914 Chr.17: 35750023-35750023 on Build GRCh38
rs782185235 Chr.17: 35750107-35750107 on Build GRCh38
rs998384866 Chr.17: 35749972-35749972 on Build GRCh38
rs782170313 Chr.17: 35750050-35750050 on Build GRCh38
rs782223631 Chr.17: 35750026-35750026 on Build GRCh38
rs370646887 Chr.17: 35750081-35750081 on Build GRCh38
rs782426226 Chr.17: 35750108-35750108 on Build GRCh38

View Full Product Details