Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109816152-109816423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAR1
Entrez Gene ID
54433
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
NOLA1
UniGene
Hs.69851
Gene Chromosome Location
Chr.4: 109815510-109824737 on Build GRCh38
Transcripts
NM_018983.3 NM_032993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6325739 c.152G>T p.R51L Chr.4: 109816316-109816316 on Build GRCh38
447228 c.31T>G p.F11V Chr.4: 109816195-109816195 on Build GRCh38
3006249 c.158G>A p.G53D Chr.4: 109816322-109816322 on Build GRCh38
3006250 c.193G>T p.G65* Chr.4: 109816357-109816357 on Build GRCh38
480744 c.10C>T p.R4* Chr.4: 109816174-109816174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201161727 Chr.4: 109816321-109816321 on Build GRCh38
rs6828254 Chr.4: 109816393-109816393 on Build GRCh38
rs147082071 Chr.4: 109816247-109816247 on Build GRCh38
rs202241831 Chr.4: 109816306-109816306 on Build GRCh38
rs762679702 Chr.4: 109816258-109816260 on Build GRCh38
rs769669396 Chr.4: 109816395-109816395 on Build GRCh38
rs760555198 Chr.4: 109816183-109816183 on Build GRCh38
rs201651562 Chr.4: 109816221-109816221 on Build GRCh38
rs763131345 Chr.4: 109816207-109816207 on Build GRCh38
rs985669774 Chr.4: 109816343-109816343 on Build GRCh38
rs746751617 Chr.4: 109816282-109816282 on Build GRCh38
rs150273267 Chr.4: 109816187-109816187 on Build GRCh38
rs749898699 Chr.4: 109816335-109816335 on Build GRCh38
rs768906897 Chr.4: 109816327-109816327 on Build GRCh38
rs765203160 Chr.4: 109816255-109816266 on Build GRCh38
rs977206709 Chr.4: 109816193-109816193 on Build GRCh38
rs201487663 Chr.4: 109816377-109816377 on Build GRCh38
rs566900761 Chr.4: 109816184-109816184 on Build GRCh38
rs760010270 Chr.4: 109816323-109816323 on Build GRCh38
rs775318667 Chr.4: 109816180-109816180 on Build GRCh38
rs1016080859 Chr.4: 109816275-109816275 on Build GRCh38
rs554365787 Chr.4: 109816236-109816236 on Build GRCh38
rs753135586 Chr.4: 109816244-109816244 on Build GRCh38
rs766000039 Chr.4: 109816271-109816288 on Build GRCh38
rs368527218 Chr.4: 109816333-109816333 on Build GRCh38
rs773235585 Chr.4: 109816307-109816307 on Build GRCh38
rs779727875 Chr.4: 109816276-109816276 on Build GRCh38
rs911721637 Chr.4: 109816176-109816176 on Build GRCh38
rs773024250 Chr.4: 109816254-109816255 on Build GRCh38
rs756651159 Chr.4: 109816329-109816329 on Build GRCh38
rs746629925 Chr.4: 109816365-109816365 on Build GRCh38
rs748054066 Chr.4: 109816297-109816297 on Build GRCh38
rs779823331 Chr.4: 109816354-109816354 on Build GRCh38
rs768334386 Chr.4: 109816294-109816294 on Build GRCh38
rs767757484 Chr.4: 109816324-109816324 on Build GRCh38
rs774579065 Chr.4: 109816322-109816322 on Build GRCh38
rs758077461 Chr.4: 109816268-109816268 on Build GRCh38
rs61737734 Chr.4: 109816296-109816296 on Build GRCh38
rs376764422 Chr.4: 109816330-109816330 on Build GRCh38
rs753291060 Chr.4: 109816325-109816325 on Build GRCh38
rs149585427 Chr.4: 109816301-109816301 on Build GRCh38
rs745549880 Chr.4: 109816256-109816256 on Build GRCh38
rs995441741 Chr.4: 109816352-109816352 on Build GRCh38
rs2276326 Chr.4: 109816233-109816233 on Build GRCh38
rs377146693 Chr.4: 109816345-109816345 on Build GRCh38
rs751178188 Chr.4: 109816270-109816271 on Build GRCh38
rs776752804 Chr.4: 109816209-109816210 on Build GRCh38
rs766758866 Chr.4: 109816218-109816218 on Build GRCh38
rs750692346 Chr.4: 109816204-109816204 on Build GRCh38
rs1028744958 Chr.4: 109816384-109816384 on Build GRCh38
rs147947279 Chr.4: 109816203-109816203 on Build GRCh38
rs780817216 Chr.4: 109816387-109816387 on Build GRCh38

View Full Product Details