Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110163374-110163647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A1
Entrez Gene ID
1282
Gene Name
collagen type IV alpha 1 chain
Gene Aliases
BSVD, RATOR
UniGene
Hs.17441
Gene Chromosome Location
Chr.13: 110148958-110307157 on Build GRCh38
Transcripts
NM_001845.5 XM_011521048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707010 c.3121C>T p.P1041S Chr.13: 110163520-110163520 on Build GRCh38
696040 c.4204G>T p.G1402C Chr.13: 110163508-110163508 on Build GRCh38
4045917 c.4238C>A p.A1413D Chr.13: 110163474-110163474 on Build GRCh38
3467110 c.3154G>A p.E1052K Chr.13: 110163487-110163487 on Build GRCh38
696042 c.4240G>T p.G1414W Chr.13: 110163472-110163472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746699679 Chr.13: 110163611-110163611 on Build GRCh38
rs182728431 Chr.13: 110163545-110163545 on Build GRCh38
rs944085788 Chr.13: 110163598-110163598 on Build GRCh38
rs200734000 Chr.13: 110163483-110163483 on Build GRCh38
rs1040074177 Chr.13: 110163600-110163600 on Build GRCh38
rs150594280 Chr.13: 110163570-110163570 on Build GRCh38
rs141714960 Chr.13: 110163509-110163509 on Build GRCh38
rs758563901 Chr.13: 110163415-110163415 on Build GRCh38
rs1010636679 Chr.13: 110163426-110163426 on Build GRCh38
rs778439115 Chr.13: 110163446-110163446 on Build GRCh38
rs190543628 Chr.13: 110163471-110163471 on Build GRCh38
rs760532152 Chr.13: 110163462-110163462 on Build GRCh38
rs765105720 Chr.13: 110163487-110163487 on Build GRCh38
rs914764279 Chr.13: 110163538-110163538 on Build GRCh38
rs894794644 Chr.13: 110163411-110163411 on Build GRCh38
rs767886345 Chr.13: 110163559-110163559 on Build GRCh38
rs551567717 Chr.13: 110163435-110163435 on Build GRCh38
rs112561721 Chr.13: 110163581-110163581 on Build GRCh38
rs372976488 Chr.13: 110163491-110163491 on Build GRCh38
rs766580638 Chr.13: 110163524-110163524 on Build GRCh38
rs372602087 Chr.13: 110163432-110163432 on Build GRCh38
rs376224472 Chr.13: 110163413-110163413 on Build GRCh38
rs186028417 Chr.13: 110163454-110163454 on Build GRCh38
rs755397451 Chr.13: 110163437-110163437 on Build GRCh38
rs763020830 Chr.13: 110163522-110163522 on Build GRCh38
rs370943043 Chr.13: 110163442-110163442 on Build GRCh38
rs756588136 Chr.13: 110163571-110163571 on Build GRCh38
rs368879632 Chr.13: 110163473-110163473 on Build GRCh38
rs374682248 Chr.13: 110163457-110163457 on Build GRCh38
rs369407797 Chr.13: 110163596-110163596 on Build GRCh38
rs891848851 Chr.13: 110163591-110163605 on Build GRCh38
rs538113369 Chr.13: 110163404-110163404 on Build GRCh38
rs745368701 Chr.13: 110163590-110163590 on Build GRCh38
rs985429619 Chr.13: 110163595-110163595 on Build GRCh38
rs1004640402 Chr.13: 110163453-110163453 on Build GRCh38
rs755309537 Chr.13: 110163557-110163557 on Build GRCh38
rs368150613 Chr.13: 110163478-110163478 on Build GRCh38
rs183228479 Chr.13: 110163441-110163441 on Build GRCh38
rs1015139357 Chr.13: 110163470-110163470 on Build GRCh38

View Full Product Details