Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13801257-13801486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUR1
Entrez Gene ID
63933
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
C6orf79, CCDC90A, FMP32
UniGene
Hs.214043
Gene Chromosome Location
Chr.6: 13786549-13814994 on Build GRCh38
Transcripts
NM_001031713.3 XM_011514802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3015670 c.686delA p.K229fs*3 Chr.6: 13801343-13801343 on Build GRCh38
6710147 c.691A>G p.M231V Chr.6: 13801338-13801338 on Build GRCh38
1073796 c.674C>T p.A225V Chr.6: 13801355-13801355 on Build GRCh38
5450188 c.640-1G>T p.? Chr.6: 13801390-13801390 on Build GRCh38
150055 c.646A>G p.T216A Chr.6: 13801383-13801383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372541722 Chr.6: 13801341-13801341 on Build GRCh38
rs776808372 Chr.6: 13801441-13801441 on Build GRCh38
rs767442278 Chr.6: 13801296-13801296 on Build GRCh38
rs748819315 Chr.6: 13801371-13801371 on Build GRCh38
rs763153046 Chr.6: 13801334-13801336 on Build GRCh38
rs3173478 Chr.6: 13801360-13801360 on Build GRCh38
rs766515581 Chr.6: 13801314-13801314 on Build GRCh38
rs759646202 Chr.6: 13801416-13801416 on Build GRCh38
rs894915360 Chr.6: 13801362-13801362 on Build GRCh38
rs759699139 Chr.6: 13801331-13801331 on Build GRCh38
rs1052422518 Chr.6: 13801413-13801413 on Build GRCh38
rs764346845 Chr.6: 13801415-13801415 on Build GRCh38
rs768555854 Chr.6: 13801285-13801285 on Build GRCh38
rs1016585041 Chr.6: 13801390-13801390 on Build GRCh38
rs760715136 Chr.6: 13801307-13801307 on Build GRCh38
rs747955303 Chr.6: 13801382-13801382 on Build GRCh38
rs776628134 Chr.6: 13801415-13801415 on Build GRCh38
rs765448670 Chr.6: 13801440-13801440 on Build GRCh38
rs921818197 Chr.6: 13801322-13801322 on Build GRCh38
rs768355916 Chr.6: 13801374-13801374 on Build GRCh38
rs761545857 Chr.6: 13801293-13801293 on Build GRCh38
rs969643031 Chr.6: 13801450-13801450 on Build GRCh38
rs201100064 Chr.6: 13801342-13801342 on Build GRCh38
rs778454712 Chr.6: 13801376-13801376 on Build GRCh38
rs375249304 Chr.6: 13801316-13801316 on Build GRCh38
rs145895773 Chr.6: 13801355-13801355 on Build GRCh38
rs141962734 Chr.6: 13801354-13801354 on Build GRCh38
rs765452569 Chr.6: 13801332-13801332 on Build GRCh38
rs955229065 Chr.6: 13801414-13801418 on Build GRCh38
rs770872708 Chr.6: 13801401-13801401 on Build GRCh38
rs773198097 Chr.6: 13801387-13801387 on Build GRCh38
rs571202636 Chr.6: 13801446-13801446 on Build GRCh38
rs773072948 Chr.6: 13801298-13801298 on Build GRCh38
rs755680560 Chr.6: 13801357-13801357 on Build GRCh38
rs994048221 Chr.6: 13801409-13801409 on Build GRCh38
rs369239007 Chr.6: 13801340-13801340 on Build GRCh38
rs3180196 Chr.6: 13801383-13801383 on Build GRCh38
rs760667816 Chr.6: 13801392-13801392 on Build GRCh38
rs138786106 Chr.6: 13801288-13801288 on Build GRCh38

View Full Product Details