Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29064862-29065095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf71
Entrez Gene ID
388939
Gene Name
chromosome 2 open reading frame 71
Gene Aliases
RP54
UniGene
Hs.354243
Gene Chromosome Location
Chr.2: 29061690-29074261 on Build GRCh38
Transcripts
NM_001029883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358503 c.3725C>A p.P1242H Chr.2: 29065011-29065011 on Build GRCh38
319079 c.3767T>C p.F1256S Chr.2: 29064969-29064969 on Build GRCh38
6415115 c.3865T>A p.*1289R Chr.2: 29064871-29064871 on Build GRCh38
3972217 c.3839C>T p.A1280V Chr.2: 29064897-29064897 on Build GRCh38
3580930 c.3828C>T p.S1276S Chr.2: 29064908-29064908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376456208 Chr.2: 29064933-29064933 on Build GRCh38
rs201767517 Chr.2: 29064910-29064910 on Build GRCh38
rs779792135 Chr.2: 29064964-29064964 on Build GRCh38
rs781712767 Chr.2: 29064900-29064900 on Build GRCh38
rs775287807 Chr.2: 29064934-29064934 on Build GRCh38
rs370823165 Chr.2: 29065006-29065006 on Build GRCh38
rs572507416 Chr.2: 29064956-29064956 on Build GRCh38
rs760408273 Chr.2: 29064936-29064936 on Build GRCh38
rs199853956 Chr.2: 29065028-29065028 on Build GRCh38
rs770436734 Chr.2: 29064906-29064906 on Build GRCh38
rs1031601220 Chr.2: 29064962-29064962 on Build GRCh38
rs748471967 Chr.2: 29064901-29064901 on Build GRCh38
rs748527009 Chr.2: 29064973-29064973 on Build GRCh38
rs886055920 Chr.2: 29065072-29065072 on Build GRCh38
rs779257474 Chr.2: 29065063-29065064 on Build GRCh38
rs115730331 Chr.2: 29064896-29064896 on Build GRCh38
rs59833221 Chr.2: 29065071-29065072 on Build GRCh38
rs764839767 Chr.2: 29064952-29064952 on Build GRCh38
rs201566996 Chr.2: 29064953-29064953 on Build GRCh38
rs778285450 Chr.2: 29064888-29064888 on Build GRCh38
rs201781577 Chr.2: 29065060-29065060 on Build GRCh38
rs536011264 Chr.2: 29065058-29065058 on Build GRCh38
rs771588718 Chr.2: 29064897-29064897 on Build GRCh38
rs954144109 Chr.2: 29064998-29064998 on Build GRCh38
rs727503829 Chr.2: 29064988-29064988 on Build GRCh38
rs963611260 Chr.2: 29064930-29064930 on Build GRCh38
rs139768554 Chr.2: 29065060-29065061 on Build GRCh38
rs187333111 Chr.2: 29064997-29064997 on Build GRCh38
rs756746994 Chr.2: 29065028-29065028 on Build GRCh38
rs867375433 Chr.2: 29065005-29065005 on Build GRCh38
rs369699375 Chr.2: 29064941-29064941 on Build GRCh38
rs78471512 Chr.2: 29065014-29065014 on Build GRCh38
rs576465020 Chr.2: 29064929-29064929 on Build GRCh38
rs749848766 Chr.2: 29064987-29064987 on Build GRCh38
rs191767954 Chr.2: 29065031-29065031 on Build GRCh38
rs368860499 Chr.2: 29065001-29065001 on Build GRCh38
rs745549188 Chr.2: 29065061-29065063 on Build GRCh38
rs564406283 Chr.2: 29065072-29065073 on Build GRCh38
rs928963573 Chr.2: 29065072-29065072 on Build GRCh38
rs200738006 Chr.2: 29064977-29064977 on Build GRCh38
rs199689791 Chr.2: 29064947-29064947 on Build GRCh38
rs386389811 Chr.2: 29065069-29065070 on Build GRCh38
rs757355249 Chr.2: 29065066-29065067 on Build GRCh38
rs369662857 Chr.2: 29064923-29064923 on Build GRCh38
rs772297494 Chr.2: 29065004-29065004 on Build GRCh38
rs373543146 Chr.2: 29064928-29064928 on Build GRCh38
rs753843724 Chr.2: 29064942-29064942 on Build GRCh38
rs749808562 Chr.2: 29064920-29064920 on Build GRCh38
rs1975713 Chr.2: 29064975-29064975 on Build GRCh38
rs886042981 Chr.2: 29064958-29064958 on Build GRCh38
rs368652431 Chr.2: 29065022-29065022 on Build GRCh38
rs201933656 Chr.2: 29065032-29065032 on Build GRCh38

View Full Product Details