Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2898938-2899201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLYWCH2
Entrez Gene ID
114984
Gene Name
FLYWCH family member 2
Gene Aliases
-
UniGene
Hs.534525
Gene Chromosome Location
Chr.16: 2882891-2899382 on Build GRCh38
Transcripts
NM_001142499.1 NM_001142500.1 NM_138439.2 XM_005255078.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6221710 c.365_369delCTGAG p.P122fs*>18 Chr.16: 2899091-2899095 on Build GRCh38
6665413 c.349T>C p.L117L Chr.16: 2899075-2899075 on Build GRCh38
1377217 c.363T>C p.P121P Chr.16: 2899089-2899089 on Build GRCh38
6665419 c.388_389insC p.C132fs*>10 Chr.16: 2899114-2899115 on Build GRCh38
118349 c.384C>T p.N128N Chr.16: 2899110-2899110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141682013 Chr.16: 2899134-2899134 on Build GRCh38
rs143698118 Chr.16: 2899148-2899148 on Build GRCh38
rs377269240 Chr.16: 2899002-2899002 on Build GRCh38
rs773639666 Chr.16: 2899156-2899156 on Build GRCh38
rs992072978 Chr.16: 2899057-2899057 on Build GRCh38
rs528311318 Chr.16: 2898986-2898986 on Build GRCh38
rs945185289 Chr.16: 2899060-2899060 on Build GRCh38
rs145106450 Chr.16: 2899070-2899070 on Build GRCh38
rs373608135 Chr.16: 2899165-2899165 on Build GRCh38
rs111514958 Chr.16: 2899164-2899164 on Build GRCh38
rs1008451387 Chr.16: 2898985-2898985 on Build GRCh38
rs753626721 Chr.16: 2899124-2899124 on Build GRCh38
rs754366085 Chr.16: 2899000-2899000 on Build GRCh38
rs373664446 Chr.16: 2899083-2899083 on Build GRCh38
rs926278418 Chr.16: 2899046-2899046 on Build GRCh38
rs370800734 Chr.16: 2899014-2899014 on Build GRCh38
rs776186024 Chr.16: 2899120-2899120 on Build GRCh38
rs555954202 Chr.16: 2899115-2899115 on Build GRCh38
rs369626075 Chr.16: 2899096-2899096 on Build GRCh38
rs758290758 Chr.16: 2899175-2899175 on Build GRCh38
rs548440333 Chr.16: 2898994-2898994 on Build GRCh38
rs759007390 Chr.16: 2899107-2899107 on Build GRCh38
rs375468185 Chr.16: 2899001-2899001 on Build GRCh38
rs754497244 Chr.16: 2899004-2899004 on Build GRCh38
rs960678949 Chr.16: 2899055-2899055 on Build GRCh38
rs761720578 Chr.16: 2899128-2899134 on Build GRCh38
rs755784897 Chr.16: 2899117-2899117 on Build GRCh38
rs780013780 Chr.16: 2899178-2899178 on Build GRCh38
rs557821707 Chr.16: 2899135-2899135 on Build GRCh38
rs372331570 Chr.16: 2899062-2899062 on Build GRCh38
rs201098556 Chr.16: 2899140-2899140 on Build GRCh38
rs371986596 Chr.16: 2899163-2899163 on Build GRCh38
rs7204314 Chr.16: 2899174-2899174 on Build GRCh38
rs757171625 Chr.16: 2899168-2899168 on Build GRCh38
rs199839518 Chr.16: 2899044-2899044 on Build GRCh38
rs780774184 Chr.16: 2899012-2899012 on Build GRCh38
rs34724597 Chr.16: 2899095-2899096 on Build GRCh38
rs377448745 Chr.16: 2899170-2899170 on Build GRCh38
rs149108052 Chr.16: 2899121-2899121 on Build GRCh38
rs200210861 Chr.16: 2899018-2899018 on Build GRCh38
rs749267388 Chr.16: 2899022-2899022 on Build GRCh38
rs369874426 Chr.16: 2899131-2899131 on Build GRCh38
rs376595971 Chr.16: 2899130-2899130 on Build GRCh38
rs550299892 Chr.16: 2899025-2899025 on Build GRCh38
rs757118369 Chr.16: 2899126-2899126 on Build GRCh38
rs374160385 Chr.16: 2899108-2899108 on Build GRCh38
rs146456984 Chr.16: 2899086-2899086 on Build GRCh38
rs951758838 Chr.16: 2899029-2899029 on Build GRCh38
rs750935692 Chr.16: 2899092-2899092 on Build GRCh38
rs763467628 Chr.16: 2899159-2899159 on Build GRCh38
rs748579032 Chr.16: 2899143-2899143 on Build GRCh38
rs201846710 Chr.16: 2899039-2899039 on Build GRCh38
rs201531918 Chr.16: 2899045-2899045 on Build GRCh38
rs776916629 Chr.16: 2899080-2899080 on Build GRCh38
rs765034719 Chr.16: 2899173-2899173 on Build GRCh38
rs545946886 Chr.16: 2899181-2899181 on Build GRCh38
rs190439473 Chr.16: 2899021-2899021 on Build GRCh38
rs913849606 Chr.16: 2899059-2899059 on Build GRCh38
rs566687400 Chr.16: 2899082-2899082 on Build GRCh38
rs772165323 Chr.16: 2899042-2899042 on Build GRCh38
rs1016289711 Chr.16: 2898990-2898990 on Build GRCh38
rs891140124 Chr.16: 2898962-2898962 on Build GRCh38
rs1008534715 Chr.16: 2899087-2899087 on Build GRCh38
rs766964544 Chr.16: 2899162-2899162 on Build GRCh38

View Full Product Details