Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132453971-132454230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616925 c.932+4delT p.? Chr.3: 132454161-132454161 on Build GRCh38
237309 c.906A>G p.Q302Q Chr.3: 132454131-132454131 on Build GRCh38
4846732 c.928G>C p.E310Q Chr.3: 132454153-132454153 on Build GRCh38
6219655 c.904C>T p.Q302* Chr.3: 132454129-132454129 on Build GRCh38
1038398 c.842T>C p.V281A Chr.3: 132454067-132454067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141158901 Chr.3: 132454096-132454096 on Build GRCh38
rs764501989 Chr.3: 132454126-132454126 on Build GRCh38
rs147315244 Chr.3: 132454135-132454135 on Build GRCh38
rs372419664 Chr.3: 132454098-132454098 on Build GRCh38
rs767766724 Chr.3: 132454173-132454173 on Build GRCh38
rs770229103 Chr.3: 132454114-132454114 on Build GRCh38
rs749068099 Chr.3: 132454068-132454068 on Build GRCh38
rs762125039 Chr.3: 132454143-132454143 on Build GRCh38
rs368485440 Chr.3: 132454074-132454074 on Build GRCh38
rs555862676 Chr.3: 132454112-132454112 on Build GRCh38
rs753761171 Chr.3: 132454200-132454200 on Build GRCh38
rs755981031 Chr.3: 132454037-132454037 on Build GRCh38
rs776028296 Chr.3: 132454116-132454116 on Build GRCh38
rs751769772 Chr.3: 132454025-132454025 on Build GRCh38
rs200602631 Chr.3: 132454190-132454190 on Build GRCh38
rs761395737 Chr.3: 132454030-132454030 on Build GRCh38
rs185502283 Chr.3: 132454208-132454208 on Build GRCh38
rs756154616 Chr.3: 132454192-132454192 on Build GRCh38
rs57786343 Chr.3: 132454173-132454174 on Build GRCh38
rs755532444 Chr.3: 132454029-132454029 on Build GRCh38
rs866905371 Chr.3: 132454046-132454046 on Build GRCh38
rs750546597 Chr.3: 132454191-132454191 on Build GRCh38
rs747862340 Chr.3: 132454205-132454205 on Build GRCh38
rs542534222 Chr.3: 132454161-132454161 on Build GRCh38
rs771637349 Chr.3: 132454084-132454084 on Build GRCh38
rs778646742 Chr.3: 132454203-132454203 on Build GRCh38
rs139147920 Chr.3: 132454021-132454021 on Build GRCh38
rs371902534 Chr.3: 132454014-132454014 on Build GRCh38
rs757465755 Chr.3: 132454026-132454026 on Build GRCh38
rs766259792 Chr.3: 132454196-132454196 on Build GRCh38
rs755845431 Chr.3: 132454160-132454161 on Build GRCh38
rs1055718366 Chr.3: 132454163-132454163 on Build GRCh38
rs544590152 Chr.3: 132454202-132454202 on Build GRCh38
rs781733584 Chr.3: 132454161-132454162 on Build GRCh38
rs779990962 Chr.3: 132454049-132454049 on Build GRCh38
rs888682753 Chr.3: 132454120-132454120 on Build GRCh38
rs747785821 Chr.3: 132454077-132454077 on Build GRCh38
rs781413600 Chr.3: 132454028-132454028 on Build GRCh38
rs770515952 Chr.3: 132454200-132454211 on Build GRCh38
rs750423757 Chr.3: 132454032-132454032 on Build GRCh38
rs763254633 Chr.3: 132454117-132454117 on Build GRCh38
rs768430183 Chr.3: 132454073-132454073 on Build GRCh38

View Full Product Details