Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11104382-11104639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVFRD-1
Entrez Gene ID
405754
Gene Name
endogenous retrovirus group FRD member 1
Gene Aliases
ERVFRDE1, GLLL6191, HERV-FRD, HERV-W/FRD, UNQ6191, envFRD
UniGene
Hs.631996
Gene Chromosome Location
Chr.6: 11102489-11111838 on Build GRCh38
Transcripts
NM_207582.2 NM_001135575.1

Gene Symbol
SMIM13
Entrez Gene ID
221710
Gene Name
small integral membrane protein 13
Gene Aliases
C6orf228
UniGene
Hs.656600
Gene Chromosome Location
Chr.6: 11094033-11138738 on Build GRCh38
Transcripts
NM_207582.2 NM_001135575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150050 c.923G>T p.C308F Chr.6: 11104388-11104388 on Build GRCh38
1072153 c.889G>T p.A297S Chr.6: 11104422-11104422 on Build GRCh38
7366599 c.887G>C p.G296A Chr.6: 11104424-11104424 on Build GRCh38
6036091 c.844C>A p.P282T Chr.6: 11104467-11104467 on Build GRCh38
5452070 c.828T>C p.F276F Chr.6: 11104483-11104483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184878901 Chr.6: 11104441-11104441 on Build GRCh38
rs752839265 Chr.6: 11104484-11104484 on Build GRCh38
rs145616782 Chr.6: 11104447-11104447 on Build GRCh38
rs756366726 Chr.6: 11104506-11104506 on Build GRCh38
rs150256083 Chr.6: 11104559-11104559 on Build GRCh38
rs780448839 Chr.6: 11104507-11104507 on Build GRCh38
rs768912764 Chr.6: 11104550-11104550 on Build GRCh38
rs369585109 Chr.6: 11104608-11104608 on Build GRCh38
rs760745944 Chr.6: 11104541-11104541 on Build GRCh38
rs772643136 Chr.6: 11104551-11104551 on Build GRCh38
rs776050141 Chr.6: 11104552-11104552 on Build GRCh38
rs72825119 Chr.6: 11104505-11104505 on Build GRCh38
rs760124620 Chr.6: 11104421-11104421 on Build GRCh38
rs558720062 Chr.6: 11104577-11104577 on Build GRCh38
rs745426977 Chr.6: 11104511-11104511 on Build GRCh38
rs1045562078 Chr.6: 11104459-11104459 on Build GRCh38
rs749572811 Chr.6: 11104512-11104512 on Build GRCh38
rs773537004 Chr.6: 11104577-11104577 on Build GRCh38
rs1023869297 Chr.6: 11104461-11104461 on Build GRCh38
rs114277097 Chr.6: 11104414-11104414 on Build GRCh38
rs767522278 Chr.6: 11104583-11104583 on Build GRCh38
rs779404885 Chr.6: 11104526-11104526 on Build GRCh38
rs557053848 Chr.6: 11104406-11104406 on Build GRCh38
rs754111903 Chr.6: 11104611-11104611 on Build GRCh38
rs759075554 Chr.6: 11104554-11104554 on Build GRCh38
rs763998164 Chr.6: 11104424-11104424 on Build GRCh38
rs761589046 Chr.6: 11104582-11104582 on Build GRCh38
rs763792082 Chr.6: 11104572-11104572 on Build GRCh38
rs768876073 Chr.6: 11104517-11104517 on Build GRCh38
rs781074920 Chr.6: 11104502-11104502 on Build GRCh38
rs747427247 Chr.6: 11104595-11104596 on Build GRCh38
rs769386245 Chr.6: 11104557-11104557 on Build GRCh38
rs756242354 Chr.6: 11104610-11104610 on Build GRCh38

View Full Product Details