Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3256027-3256256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf194
Entrez Gene ID
25943
Gene Name
chromosome 20 open reading frame 194
Gene Aliases
-
UniGene
Hs.516853
Gene Chromosome Location
Chr.20: 3249302-3407662 on Build GRCh38
Transcripts
NM_001009984.2 XM_005260684.3 XM_005260687.4 XM_011529208.2 XM_017027783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4097742 c.3157G>A p.V1053I Chr.20: 3256110-3256110 on Build GRCh38
4097741 c.3232A>G p.K1078E Chr.20: 3256035-3256035 on Build GRCh38
6313044 c.3237C>A p.D1079E Chr.20: 3256030-3256030 on Build GRCh38
6419335 c.3227G>T p.S1076I Chr.20: 3256040-3256040 on Build GRCh38
6615519 c.3204C>T p.I1068I Chr.20: 3256063-3256063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922905225 Chr.20: 3256192-3256192 on Build GRCh38
rs375035389 Chr.20: 3256155-3256155 on Build GRCh38
rs758341025 Chr.20: 3256137-3256137 on Build GRCh38
rs979859122 Chr.20: 3256228-3256229 on Build GRCh38
rs776190544 Chr.20: 3256122-3256122 on Build GRCh38
rs765001505 Chr.20: 3256125-3256125 on Build GRCh38
rs140305967 Chr.20: 3256110-3256110 on Build GRCh38
rs779184374 Chr.20: 3256162-3256162 on Build GRCh38
rs780158228 Chr.20: 3256092-3256092 on Build GRCh38
rs961987160 Chr.20: 3256140-3256140 on Build GRCh38
rs772519155 Chr.20: 3256193-3256193 on Build GRCh38
rs746500208 Chr.20: 3256216-3256216 on Build GRCh38
rs913582913 Chr.20: 3256230-3256230 on Build GRCh38
rs768941591 Chr.20: 3256096-3256096 on Build GRCh38
rs199661270 Chr.20: 3256121-3256121 on Build GRCh38
rs752556551 Chr.20: 3256127-3256127 on Build GRCh38
rs369655093 Chr.20: 3256081-3256081 on Build GRCh38
rs749641626 Chr.20: 3256093-3256093 on Build GRCh38
rs1050112168 Chr.20: 3256225-3256225 on Build GRCh38
rs926400821 Chr.20: 3256157-3256157 on Build GRCh38
rs753621233 Chr.20: 3256062-3256062 on Build GRCh38
rs765194129 Chr.20: 3256065-3256065 on Build GRCh38
rs774177629 Chr.20: 3256230-3256231 on Build GRCh38
rs1035392900 Chr.20: 3256084-3256084 on Build GRCh38
rs772313170 Chr.20: 3256078-3256078 on Build GRCh38
rs201873954 Chr.20: 3256083-3256083 on Build GRCh38
rs776929308 Chr.20: 3256221-3256221 on Build GRCh38
rs953466471 Chr.20: 3256120-3256120 on Build GRCh38
rs748544734 Chr.20: 3256189-3256189 on Build GRCh38
rs552252849 Chr.20: 3256112-3256112 on Build GRCh38
rs936452508 Chr.20: 3256213-3256213 on Build GRCh38
rs754781920 Chr.20: 3256063-3256063 on Build GRCh38
rs371119992 Chr.20: 3256148-3256148 on Build GRCh38
rs200570338 Chr.20: 3256152-3256152 on Build GRCh38
rs760125786 Chr.20: 3256117-3256117 on Build GRCh38
rs772670526 Chr.20: 3256115-3256115 on Build GRCh38
rs768320154 Chr.20: 3256229-3256230 on Build GRCh38
rs200794835 Chr.20: 3256057-3256057 on Build GRCh38
rs752752371 Chr.20: 3256077-3256077 on Build GRCh38
rs200104613 Chr.20: 3256055-3256055 on Build GRCh38
rs375669005 Chr.20: 3256123-3256123 on Build GRCh38
rs377454144 Chr.20: 3256153-3256153 on Build GRCh38
rs201711364 Chr.20: 3256111-3256111 on Build GRCh38
rs755355925 Chr.20: 3256160-3256160 on Build GRCh38
rs375288538 Chr.20: 3256087-3256087 on Build GRCh38
rs773757507 Chr.20: 3256201-3256201 on Build GRCh38

View Full Product Details