Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102603076-102603331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5
Entrez Gene ID
64860
Gene Name
armadillo repeat containing, X-linked 5
Gene Aliases
GASP5
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599168-102604159 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001168478.1 NM_001168479.1 NM_001168480.1 NM_001168482.1 NM_001168485.1 NM_022838.3 NM_001199818.1

SNP Detail

SNP ID Chromosome Location
rs781341075 Chr.X: 102603167-102603167 on Build GRCh38
rs746711877 Chr.X: 102603235-102603235 on Build GRCh38
rs757444846 Chr.X: 102603164-102603164 on Build GRCh38
rs761553267 Chr.X: 102603300-102603300 on Build GRCh38
rs1000634112 Chr.X: 102603207-102603207 on Build GRCh38
rs200145435 Chr.X: 102603275-102603275 on Build GRCh38
rs750531280 Chr.X: 102603180-102603180 on Build GRCh38
rs866416226 Chr.X: 102603186-102603186 on Build GRCh38
rs749789460 Chr.X: 102603222-102603222 on Build GRCh38
rs374899022 Chr.X: 102603290-102603290 on Build GRCh38
rs776386566 Chr.X: 102603269-102603269 on Build GRCh38
rs768968357 Chr.X: 102603226-102603226 on Build GRCh38
rs1019659336 Chr.X: 102603150-102603150 on Build GRCh38
rs751377279 Chr.X: 102603299-102603299 on Build GRCh38
rs535328372 Chr.X: 102603278-102603278 on Build GRCh38
rs762914687 Chr.X: 102603291-102603291 on Build GRCh38
rs780437303 Chr.X: 102603194-102603194 on Build GRCh38
rs868738831 Chr.X: 102603148-102603148 on Build GRCh38
rs1038880909 Chr.X: 102603221-102603221 on Build GRCh38
rs767404642 Chr.X: 102603232-102603232 on Build GRCh38
rs770732927 Chr.X: 102603264-102603264 on Build GRCh38
rs752535299 Chr.X: 102603284-102603284 on Build GRCh38
rs756152594 Chr.X: 102603187-102603187 on Build GRCh38
rs763841626 Chr.X: 102603297-102603297 on Build GRCh38
rs370182133 Chr.X: 102603139-102603139 on Build GRCh38

View Full Product Details