Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52059775-52060024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3L4
Entrez Gene ID
91408
Gene Name
basic transcription factor 3 like 4
Gene Aliases
-
UniGene
Hs.429839
Gene Chromosome Location
Chr.1: 52056185-52090716 on Build GRCh38
Transcripts
NM_001136497.2 NM_001243767.1 NM_152265.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3944167 c.46G>C p.G16R Chr.1: 52059893-52059893 on Build GRCh38
910611 c.39C>T p.V13V Chr.1: 52059886-52059886 on Build GRCh38
2192722 c.22A>T p.K8* Chr.1: 52059869-52059869 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752725560 Chr.1: 52059819-52059820 on Build GRCh38
rs768164012 Chr.1: 52059913-52059913 on Build GRCh38
rs373896304 Chr.1: 52059944-52059944 on Build GRCh38
rs778763987 Chr.1: 52059957-52059957 on Build GRCh38
rs756995696 Chr.1: 52059947-52059947 on Build GRCh38
rs139241125 Chr.1: 52059805-52059805 on Build GRCh38
rs764239131 Chr.1: 52059824-52059825 on Build GRCh38
rs776492742 Chr.1: 52059827-52059827 on Build GRCh38
rs141721999 Chr.1: 52059866-52059866 on Build GRCh38
rs754059104 Chr.1: 52059825-52059825 on Build GRCh38
rs756661126 Chr.1: 52059926-52059926 on Build GRCh38
rs774583297 Chr.1: 52059881-52059881 on Build GRCh38
rs1024536746 Chr.1: 52059873-52059873 on Build GRCh38
rs887331720 Chr.1: 52059835-52059835 on Build GRCh38
rs777484964 Chr.1: 52059824-52059824 on Build GRCh38
rs750924718 Chr.1: 52059923-52059923 on Build GRCh38
rs762212506 Chr.1: 52059888-52059888 on Build GRCh38
rs756342032 Chr.1: 52059820-52059820 on Build GRCh38
rs573098344 Chr.1: 52059966-52059966 on Build GRCh38
rs759633983 Chr.1: 52059829-52059829 on Build GRCh38
rs575235266 Chr.1: 52059955-52059955 on Build GRCh38
rs377599849 Chr.1: 52059830-52059830 on Build GRCh38
rs370570180 Chr.1: 52059895-52059895 on Build GRCh38
rs536099635 Chr.1: 52059964-52059964 on Build GRCh38
rs903228638 Chr.1: 52059814-52059814 on Build GRCh38
rs10493163 Chr.1: 52059965-52059965 on Build GRCh38
rs770856329 Chr.1: 52059826-52059826 on Build GRCh38
rs764375219 Chr.1: 52059870-52059870 on Build GRCh38
rs746798879 Chr.1: 52059825-52059825 on Build GRCh38
rs757623850 Chr.1: 52059832-52059832 on Build GRCh38
rs905222831 Chr.1: 52059819-52059819 on Build GRCh38
rs986451722 Chr.1: 52059993-52059993 on Build GRCh38
rs540399514 Chr.1: 52059981-52059981 on Build GRCh38
rs771736317 Chr.1: 52059813-52059813 on Build GRCh38
rs1003529493 Chr.1: 52059823-52059823 on Build GRCh38
rs777413405 Chr.1: 52059945-52059945 on Build GRCh38
rs764706299 Chr.1: 52059936-52059936 on Build GRCh38

View Full Product Details