Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75312315-75312580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991158 c.3333T>C p.S1111S Chr.6: 75312499-75312499 on Build GRCh38
4765853 c.3291_3292insT p.T1098fs*>81 Chr.6: 75312540-75312541 on Build GRCh38
4807439 c.3278G>A p.R1093Q Chr.6: 75312554-75312554 on Build GRCh38
6876901 c.3339G>A p.R1113R Chr.6: 75312493-75312493 on Build GRCh38
1644612 c.3277C>T p.R1093* Chr.6: 75312555-75312555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754821014 Chr.6: 75312473-75312473 on Build GRCh38
rs202216440 Chr.6: 75312349-75312349 on Build GRCh38
rs9352202 Chr.6: 75312366-75312366 on Build GRCh38
rs781266735 Chr.6: 75312389-75312389 on Build GRCh38
rs775970649 Chr.6: 75312432-75312432 on Build GRCh38
rs866402503 Chr.6: 75312444-75312444 on Build GRCh38
rs770451999 Chr.6: 75312413-75312413 on Build GRCh38
rs371411192 Chr.6: 75312514-75312514 on Build GRCh38
rs779214829 Chr.6: 75312380-75312380 on Build GRCh38
rs765483274 Chr.6: 75312535-75312535 on Build GRCh38
rs759870641 Chr.6: 75312424-75312424 on Build GRCh38
rs772482521 Chr.6: 75312425-75312425 on Build GRCh38
rs753809858 Chr.6: 75312451-75312451 on Build GRCh38
rs573667318 Chr.6: 75312483-75312483 on Build GRCh38
rs147434989 Chr.6: 75312476-75312476 on Build GRCh38
rs761016712 Chr.6: 75312435-75312435 on Build GRCh38
rs774531931 Chr.6: 75312352-75312352 on Build GRCh38
rs748167295 Chr.6: 75312392-75312392 on Build GRCh38
rs200365491 Chr.6: 75312477-75312477 on Build GRCh38
rs748714654 Chr.6: 75312504-75312504 on Build GRCh38
rs776863722 Chr.6: 75312504-75312506 on Build GRCh38
rs776918835 Chr.6: 75312339-75312339 on Build GRCh38
rs777798341 Chr.6: 75312496-75312496 on Build GRCh38
rs764671310 Chr.6: 75312441-75312441 on Build GRCh38
rs778523737 Chr.6: 75312513-75312513 on Build GRCh38
rs754872141 Chr.6: 75312386-75312386 on Build GRCh38
rs772869232 Chr.6: 75312377-75312377 on Build GRCh38
rs374585988 Chr.6: 75312350-75312350 on Build GRCh38
rs9343281 Chr.6: 75312374-75312374 on Build GRCh38
rs142674783 Chr.6: 75312460-75312460 on Build GRCh38
rs868698674 Chr.6: 75312534-75312534 on Build GRCh38
rs773434761 Chr.6: 75312543-75312543 on Build GRCh38
rs148895165 Chr.6: 75312348-75312348 on Build GRCh38
rs575113227 Chr.6: 75312368-75312368 on Build GRCh38
rs1047582019 Chr.6: 75312459-75312459 on Build GRCh38
rs202184062 Chr.6: 75312475-75312475 on Build GRCh38
rs759923949 Chr.6: 75312365-75312365 on Build GRCh38
rs766142448 Chr.6: 75312398-75312400 on Build GRCh38
rs199701222 Chr.6: 75312544-75312544 on Build GRCh38
rs764142898 Chr.6: 75312373-75312373 on Build GRCh38
rs777090787 Chr.6: 75312407-75312407 on Build GRCh38
rs368466026 Chr.6: 75312382-75312382 on Build GRCh38
rs377318130 Chr.6: 75312525-75312525 on Build GRCh38
rs747420209 Chr.6: 75312517-75312517 on Build GRCh38
rs761789063 Chr.6: 75312533-75312533 on Build GRCh38
rs35227190 Chr.6: 75312455-75312455 on Build GRCh38
rs760729320 Chr.6: 75312372-75312372 on Build GRCh38
rs761980858 Chr.6: 75312454-75312454 on Build GRCh38
rs774030831 Chr.6: 75312414-75312414 on Build GRCh38
rs374887916 Chr.6: 75312516-75312516 on Build GRCh38
rs766731465 Chr.6: 75312554-75312554 on Build GRCh38
rs915922571 Chr.6: 75312401-75312401 on Build GRCh38
rs777247267 Chr.6: 75312526-75312526 on Build GRCh38
rs1022499710 Chr.6: 75312541-75312541 on Build GRCh38
rs748854672 Chr.6: 75312409-75312409 on Build GRCh38
rs764197668 Chr.6: 75312437-75312437 on Build GRCh38
rs267601125 Chr.6: 75312481-75312481 on Build GRCh38
rs769776078 Chr.6: 75312400-75312400 on Build GRCh38
rs752395530 Chr.6: 75312555-75312555 on Build GRCh38

View Full Product Details