Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218239298-218239571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC2
Entrez Gene ID
10109
Gene Name
actin related protein 2/3 complex subunit 2
Gene Aliases
ARC34, PNAS-139, PRO2446, p34-Arc
UniGene
Hs.529303
Gene Chromosome Location
Chr.2: 218217094-218254348 on Build GRCh38
Transcripts
NM_005731.3 NM_152862.2 XM_011510471.1 XM_017003113.1 XM_017003114.1 XM_017003115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157089 c.463G>C p.E155Q Chr.2: 218239398-218239398 on Build GRCh38
5354793 c.500C>T p.T167I Chr.2: 218239435-218239435 on Build GRCh38
5447445 c.464A>G p.E155G Chr.2: 218239399-218239399 on Build GRCh38
6487884 c.520G>A p.D174N Chr.2: 218239455-218239455 on Build GRCh38
6358659 c.483C>T p.V161V Chr.2: 218239418-218239418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899392132 Chr.2: 218239530-218239530 on Build GRCh38
rs933587323 Chr.2: 218239529-218239529 on Build GRCh38
rs750813171 Chr.2: 218239361-218239361 on Build GRCh38
rs201659815 Chr.2: 218239490-218239490 on Build GRCh38
rs781466543 Chr.2: 218239455-218239455 on Build GRCh38
rs763215689 Chr.2: 218239522-218239522 on Build GRCh38
rs187629837 Chr.2: 218239335-218239335 on Build GRCh38
rs112115567 Chr.2: 218239390-218239390 on Build GRCh38
rs185748932 Chr.2: 218239540-218239540 on Build GRCh38
rs755626586 Chr.2: 218239530-218239531 on Build GRCh38
rs760113642 Chr.2: 218239366-218239366 on Build GRCh38
rs377655586 Chr.2: 218239451-218239451 on Build GRCh38
rs982497560 Chr.2: 218239330-218239330 on Build GRCh38
rs111697110 Chr.2: 218239371-218239371 on Build GRCh38
rs1050678032 Chr.2: 218239544-218239544 on Build GRCh38
rs770396486 Chr.2: 218239509-218239509 on Build GRCh38
rs199635119 Chr.2: 218239386-218239386 on Build GRCh38
rs751703795 Chr.2: 218239531-218239531 on Build GRCh38
rs780961225 Chr.2: 218239516-218239516 on Build GRCh38
rs764236963 Chr.2: 218239442-218239442 on Build GRCh38
rs759997519 Chr.2: 218239518-218239518 on Build GRCh38
rs1041758581 Chr.2: 218239510-218239510 on Build GRCh38
rs776166622 Chr.2: 218239515-218239515 on Build GRCh38
rs200615756 Chr.2: 218239488-218239488 on Build GRCh38
rs780568820 Chr.2: 218239383-218239383 on Build GRCh38
rs762148052 Chr.2: 218239532-218239532 on Build GRCh38
rs757282923 Chr.2: 218239452-218239452 on Build GRCh38
rs771695197 Chr.2: 218239493-218239493 on Build GRCh38
rs747783346 Chr.2: 218239356-218239356 on Build GRCh38
rs114064399 Chr.2: 218239345-218239345 on Build GRCh38
rs754242837 Chr.2: 218239383-218239383 on Build GRCh38
rs373162620 Chr.2: 218239344-218239344 on Build GRCh38
rs970694340 Chr.2: 218239337-218239337 on Build GRCh38
rs528267781 Chr.2: 218239505-218239505 on Build GRCh38
rs753204594 Chr.2: 218239524-218239527 on Build GRCh38
rs749218714 Chr.2: 218239478-218239478 on Build GRCh38
rs193209358 Chr.2: 218239492-218239492 on Build GRCh38
rs114702968 Chr.2: 218239363-218239363 on Build GRCh38
rs765971436 Chr.2: 218239367-218239367 on Build GRCh38
rs142249772 Chr.2: 218239407-218239407 on Build GRCh38
rs367601012 Chr.2: 218239523-218239523 on Build GRCh38
rs1053762077 Chr.2: 218239380-218239380 on Build GRCh38
rs1018213083 Chr.2: 218239497-218239497 on Build GRCh38
rs746741120 Chr.2: 218239506-218239506 on Build GRCh38
rs775868649 Chr.2: 218239521-218239521 on Build GRCh38

View Full Product Details