Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112039239-112039468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLAT
Entrez Gene ID
1737
Gene Name
dihydrolipoamide S-acetyltransferase
Gene Aliases
DLTA, PDC-E2, PDCE2
UniGene
Hs.335551
Gene Chromosome Location
Chr.11: 112024814-112064278 on Build GRCh38
Transcripts
NM_001931.4 XM_011542647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197717 c.1035A>C p.P345P Chr.11: 112039303-112039303 on Build GRCh38
348334 c.979G>T p.A327S Chr.11: 112039247-112039247 on Build GRCh38
6644519 c.995C>T p.T332I Chr.11: 112039263-112039263 on Build GRCh38
1351264 c.1105G>T p.G369W Chr.11: 112039373-112039373 on Build GRCh38
3415535 c.982G>A p.A328T Chr.11: 112039250-112039250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782206862 Chr.11: 112039283-112039283 on Build GRCh38
rs368341648 Chr.11: 112039381-112039381 on Build GRCh38
rs781888297 Chr.11: 112039398-112039398 on Build GRCh38
rs782124389 Chr.11: 112039382-112039382 on Build GRCh38
rs376605723 Chr.11: 112039364-112039364 on Build GRCh38
rs781934980 Chr.11: 112039313-112039313 on Build GRCh38
rs782667970 Chr.11: 112039260-112039260 on Build GRCh38
rs372004817 Chr.11: 112039274-112039274 on Build GRCh38
rs369013217 Chr.11: 112039416-112039416 on Build GRCh38
rs782399309 Chr.11: 112039304-112039304 on Build GRCh38
rs782566151 Chr.11: 112039400-112039400 on Build GRCh38
rs782453793 Chr.11: 112039266-112039266 on Build GRCh38
rs375360504 Chr.11: 112039406-112039406 on Build GRCh38
rs587691942 Chr.11: 112039317-112039317 on Build GRCh38
rs587736661 Chr.11: 112039265-112039265 on Build GRCh38
rs782461159 Chr.11: 112039418-112039418 on Build GRCh38
rs146700813 Chr.11: 112039284-112039284 on Build GRCh38
rs78320677 Chr.11: 112039259-112039259 on Build GRCh38
rs782070173 Chr.11: 112039399-112039399 on Build GRCh38
rs955994394 Chr.11: 112039350-112039350 on Build GRCh38
rs140302942 Chr.11: 112039303-112039303 on Build GRCh38
rs781982618 Chr.11: 112039290-112039290 on Build GRCh38
rs372897026 Chr.11: 112039386-112039386 on Build GRCh38
rs982032675 Chr.11: 112039308-112039308 on Build GRCh38
rs782748917 Chr.11: 112039346-112039346 on Build GRCh38

View Full Product Details