Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73725173-73725430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5045529 c.2488T>G p.S830A Chr.14: 73725358-73725358 on Build GRCh38
4939680 c.2567A>G p.Q856R Chr.14: 73725279-73725279 on Build GRCh38
6905483 c.2497T>C p.W833R Chr.14: 73725349-73725349 on Build GRCh38
6573449 c.2556C>G p.F852L Chr.14: 73725290-73725290 on Build GRCh38
1748974 c.2565G>T p.V855V Chr.14: 73725281-73725281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112726996 Chr.14: 73725265-73725265 on Build GRCh38
rs976353141 Chr.14: 73725253-73725253 on Build GRCh38
rs865879558 Chr.14: 73725339-73725339 on Build GRCh38
rs756023163 Chr.14: 73725307-73725307 on Build GRCh38
rs932367436 Chr.14: 73725276-73725276 on Build GRCh38
rs779920233 Chr.14: 73725319-73725319 on Build GRCh38
rs200559216 Chr.14: 73725363-73725363 on Build GRCh38
rs200122114 Chr.14: 73725401-73725401 on Build GRCh38
rs777234710 Chr.14: 73725404-73725404 on Build GRCh38
rs772554353 Chr.14: 73725383-73725383 on Build GRCh38
rs768654583 Chr.14: 73725333-73725333 on Build GRCh38
rs1020113800 Chr.14: 73725245-73725245 on Build GRCh38
rs772553573 Chr.14: 73725373-73725373 on Build GRCh38
rs1049505354 Chr.14: 73725305-73725305 on Build GRCh38
rs1002784380 Chr.14: 73725263-73725263 on Build GRCh38
rs748305656 Chr.14: 73725359-73725359 on Build GRCh38
rs958708953 Chr.14: 73725343-73725343 on Build GRCh38
rs367695695 Chr.14: 73725241-73725241 on Build GRCh38
rs991994988 Chr.14: 73725396-73725396 on Build GRCh38
rs761309382 Chr.14: 73725400-73725400 on Build GRCh38
rs375694958 Chr.14: 73725261-73725261 on Build GRCh38
rs201589556 Chr.14: 73725321-73725321 on Build GRCh38
rs779206791 Chr.14: 73725338-73725338 on Build GRCh38
rs544043117 Chr.14: 73725208-73725208 on Build GRCh38
rs773414879 Chr.14: 73725375-73725375 on Build GRCh38

View Full Product Details