Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43085553-43085824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4678720 c.356T>C p.V119A Chr.13: 43085812-43085812 on Build GRCh38
4047714 c.319G>A p.A107T Chr.13: 43085775-43085775 on Build GRCh38
1366981 c.312-1G>A p.? Chr.13: 43085767-43085767 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs994428585 Chr.13: 43085733-43085733 on Build GRCh38
rs756740765 Chr.13: 43085774-43085774 on Build GRCh38
rs900460925 Chr.13: 43085683-43085683 on Build GRCh38
rs916898599 Chr.13: 43085602-43085602 on Build GRCh38
rs764267263 Chr.13: 43085750-43085750 on Build GRCh38
rs781672288 Chr.13: 43085758-43085758 on Build GRCh38
rs761278008 Chr.13: 43085720-43085720 on Build GRCh38
rs537282387 Chr.13: 43085771-43085771 on Build GRCh38
rs763160185 Chr.13: 43085748-43085748 on Build GRCh38
rs773879400 Chr.13: 43085719-43085719 on Build GRCh38
rs867307560 Chr.13: 43085768-43085768 on Build GRCh38
rs369956960 Chr.13: 43085746-43085746 on Build GRCh38
rs768331848 Chr.13: 43085793-43085793 on Build GRCh38
rs930561928 Chr.13: 43085705-43085705 on Build GRCh38
rs753692528 Chr.13: 43085744-43085744 on Build GRCh38
rs1008708636 Chr.13: 43085631-43085631 on Build GRCh38
rs367964575 Chr.13: 43085737-43085737 on Build GRCh38
rs948359842 Chr.13: 43085628-43085628 on Build GRCh38
rs1038925148 Chr.13: 43085653-43085653 on Build GRCh38
rs950472198 Chr.13: 43085772-43085772 on Build GRCh38
rs571524243 Chr.13: 43085620-43085620 on Build GRCh38
rs771996026 Chr.13: 43085724-43085724 on Build GRCh38
rs1044471164 Chr.13: 43085700-43085700 on Build GRCh38
rs886310745 Chr.13: 43085722-43085722 on Build GRCh38
rs531818240 Chr.13: 43085781-43085781 on Build GRCh38
rs1005051943 Chr.13: 43085786-43085786 on Build GRCh38
rs1042195756 Chr.13: 43085589-43085589 on Build GRCh38
rs751640255 Chr.13: 43085756-43085756 on Build GRCh38
rs749708619 Chr.13: 43085789-43085789 on Build GRCh38
rs992578575 Chr.13: 43085588-43085588 on Build GRCh38
rs920482854 Chr.13: 43085701-43085701 on Build GRCh38
rs557902743 Chr.13: 43085614-43085614 on Build GRCh38
rs760396231 Chr.13: 43085735-43085735 on Build GRCh38
rs757310008 Chr.13: 43085757-43085757 on Build GRCh38
rs773054604 Chr.13: 43085732-43085732 on Build GRCh38

View Full Product Details