Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157520922-157521163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL5
Entrez Gene ID
83416
Gene Name
Fc receptor like 5
Gene Aliases
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
UniGene
Hs.415950
Gene Chromosome Location
Chr.1: 157513377-157552520 on Build GRCh38
Transcripts
NM_001195388.1 NM_031281.2 XM_011510030.1 XM_011510031.1 XM_011510032.1 XM_011510033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882099 c.2464G>C p.D822H Chr.1: 157521068-157521068 on Build GRCh38
4956987 c.2462C>A p.A821D Chr.1: 157521070-157521070 on Build GRCh38
4809539 c.2444G>A p.G815E Chr.1: 157521088-157521088 on Build GRCh38
1319873 c.2468A>G p.N823S Chr.1: 157521064-157521064 on Build GRCh38
6058549 c.2427G>T p.L809L Chr.1: 157521105-157521105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781567709 Chr.1: 157520996-157520998 on Build GRCh38
rs376046891 Chr.1: 157520966-157520966 on Build GRCh38
rs758054238 Chr.1: 157521101-157521101 on Build GRCh38
rs572988825 Chr.1: 157520978-157520978 on Build GRCh38
rs74661163 Chr.1: 157521096-157521096 on Build GRCh38
rs748458889 Chr.1: 157521029-157521030 on Build GRCh38
rs767780880 Chr.1: 157521047-157521047 on Build GRCh38
rs756204225 Chr.1: 157521069-157521069 on Build GRCh38
rs757607693 Chr.1: 157520977-157520977 on Build GRCh38
rs773092893 Chr.1: 157521013-157521013 on Build GRCh38
rs771667658 Chr.1: 157520987-157520987 on Build GRCh38
rs752961202 Chr.1: 157521067-157521067 on Build GRCh38
rs773144044 Chr.1: 157521130-157521130 on Build GRCh38
rs746177534 Chr.1: 157521100-157521100 on Build GRCh38
rs769713737 Chr.1: 157521124-157521124 on Build GRCh38
rs768209443 Chr.1: 157520999-157520999 on Build GRCh38
rs540868124 Chr.1: 157520998-157520998 on Build GRCh38
rs776177266 Chr.1: 157521123-157521123 on Build GRCh38
rs779276501 Chr.1: 157521098-157521098 on Build GRCh38
rs779750979 Chr.1: 157521106-157521106 on Build GRCh38
rs200812718 Chr.1: 157521120-157521120 on Build GRCh38
rs770580193 Chr.1: 157521052-157521053 on Build GRCh38
rs376163642 Chr.1: 157521006-157521006 on Build GRCh38
rs199706458 Chr.1: 157521140-157521140 on Build GRCh38
rs544543420 Chr.1: 157521135-157521135 on Build GRCh38
rs776348207 Chr.1: 157520986-157520986 on Build GRCh38
rs754338427 Chr.1: 157521079-157521079 on Build GRCh38
rs199960121 Chr.1: 157521002-157521002 on Build GRCh38
rs367768883 Chr.1: 157521000-157521000 on Build GRCh38
rs147825124 Chr.1: 157521112-157521112 on Build GRCh38
rs761392473 Chr.1: 157521134-157521135 on Build GRCh38
rs762905791 Chr.1: 157521132-157521132 on Build GRCh38
rs759851274 Chr.1: 157521046-157521046 on Build GRCh38
rs969683347 Chr.1: 157521003-157521003 on Build GRCh38
rs961378557 Chr.1: 157520997-157520997 on Build GRCh38
rs773924683 Chr.1: 157521106-157521109 on Build GRCh38
rs554600950 Chr.1: 157520969-157520969 on Build GRCh38
rs770833408 Chr.1: 157521138-157521138 on Build GRCh38

View Full Product Details