Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 24156700-24157197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABYR
Entrez Gene ID
26256
Gene Name
calcium binding tyrosine phosphorylation regulated
Gene Aliases
CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2
UniGene
Hs.511983
Gene Chromosome Location
Chr.18: 24138956-24161600 on Build GRCh38
Transcripts
NM_001308231.1 NM_012189.3 NM_138643.2 NM_138644.2 NM_153768.2 NM_153769.2 NM_153770.2 XM_005258247.2 XM_017025706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388093 c.1378C>T p.H460Y Chr.18: 24156879-24156879 on Build GRCh38
5722724 c.1433G>A p.G478D Chr.18: 24156934-24156934 on Build GRCh38
4526462 c.1398G>A p.K466K Chr.18: 24156899-24156899 on Build GRCh38
1711040 c.1300G>A p.E434K Chr.18: 24156801-24156801 on Build GRCh38
4525398 c.1329G>A p.Q443Q Chr.18: 24156830-24156830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373252268 Chr.18: 24157019-24157019 on Build GRCh38
rs1048552709 Chr.18: 24157072-24157072 on Build GRCh38
rs755023127 Chr.18: 24157013-24157013 on Build GRCh38
rs760438730 Chr.18: 24156753-24156753 on Build GRCh38
rs764686258 Chr.18: 24156846-24156846 on Build GRCh38
rs757630307 Chr.18: 24156784-24156784 on Build GRCh38
rs770634093 Chr.18: 24156828-24156828 on Build GRCh38
rs773030785 Chr.18: 24156740-24156740 on Build GRCh38
rs376782122 Chr.18: 24156936-24156936 on Build GRCh38
rs752807337 Chr.18: 24157000-24157001 on Build GRCh38
rs746101998 Chr.18: 24156795-24156795 on Build GRCh38
rs776540121 Chr.18: 24156821-24156822 on Build GRCh38
rs969679056 Chr.18: 24156786-24156786 on Build GRCh38
rs771216494 Chr.18: 24156827-24156827 on Build GRCh38
rs772026581 Chr.18: 24156811-24156811 on Build GRCh38
rs776440512 Chr.18: 24156906-24156906 on Build GRCh38
rs1049682 Chr.18: 24156844-24156844 on Build GRCh38
rs981118678 Chr.18: 24156805-24156805 on Build GRCh38
rs752013606 Chr.18: 24156865-24156865 on Build GRCh38
rs200823555 Chr.18: 24156908-24156908 on Build GRCh38
rs141247688 Chr.18: 24156977-24156977 on Build GRCh38
rs545152652 Chr.18: 24156984-24156984 on Build GRCh38
rs199508229 Chr.18: 24156813-24156813 on Build GRCh38
rs750636638 Chr.18: 24156880-24156880 on Build GRCh38
rs747375509 Chr.18: 24156781-24156781 on Build GRCh38
rs867370082 Chr.18: 24157172-24157172 on Build GRCh38
rs746930516 Chr.18: 24156898-24156898 on Build GRCh38
rs766037794 Chr.18: 24156754-24156754 on Build GRCh38
rs142770673 Chr.18: 24156778-24156778 on Build GRCh38
rs888696501 Chr.18: 24156731-24156731 on Build GRCh38
rs764659355 Chr.18: 24156864-24156864 on Build GRCh38
rs150563017 Chr.18: 24156829-24156829 on Build GRCh38
rs944128366 Chr.18: 24157116-24157116 on Build GRCh38
rs765272590 Chr.18: 24156979-24156979 on Build GRCh38
rs762498046 Chr.18: 24156856-24156856 on Build GRCh38
rs941820890 Chr.18: 24156942-24156942 on Build GRCh38
rs571111938 Chr.18: 24157106-24157106 on Build GRCh38
rs36008645 Chr.18: 24156962-24156963 on Build GRCh38
rs1012424872 Chr.18: 24157103-24157103 on Build GRCh38
rs372528301 Chr.18: 24156761-24156761 on Build GRCh38
rs749370124 Chr.18: 24156888-24156888 on Build GRCh38
rs754113890 Chr.18: 24157006-24157006 on Build GRCh38
rs753499035 Chr.18: 24156759-24156759 on Build GRCh38
rs777695895 Chr.18: 24156890-24156890 on Build GRCh38
rs1045697191 Chr.18: 24157130-24157130 on Build GRCh38
rs190921762 Chr.18: 24156772-24156772 on Build GRCh38
rs139547401 Chr.18: 24156849-24156849 on Build GRCh38
rs184016984 Chr.18: 24156902-24156902 on Build GRCh38
rs1049683 Chr.18: 24156969-24156969 on Build GRCh38
rs775188746 Chr.18: 24157086-24157086 on Build GRCh38
rs760511204 Chr.18: 24156823-24156823 on Build GRCh38
rs562810639 Chr.18: 24156923-24156923 on Build GRCh38
rs781433714 Chr.18: 24156878-24156878 on Build GRCh38
rs756608258 Chr.18: 24156985-24156985 on Build GRCh38
rs780422361 Chr.18: 24156996-24156996 on Build GRCh38
rs759521438 Chr.18: 24156948-24156948 on Build GRCh38
rs757585802 Chr.18: 24156877-24156877 on Build GRCh38
rs759206332 Chr.18: 24156840-24156840 on Build GRCh38
rs767181111 Chr.18: 24156736-24156736 on Build GRCh38
rs781410233 Chr.18: 24156791-24156791 on Build GRCh38
rs759364155 Chr.18: 24156907-24156907 on Build GRCh38
rs533768240 Chr.18: 24157107-24157107 on Build GRCh38
rs930095147 Chr.18: 24157046-24157046 on Build GRCh38
rs777071042 Chr.18: 24156889-24156889 on Build GRCh38
rs139505147 Chr.18: 24156853-24156853 on Build GRCh38
rs985744579 Chr.18: 24156903-24156903 on Build GRCh38
rs770830634 Chr.18: 24156741-24156742 on Build GRCh38
rs142548654 Chr.18: 24156764-24156764 on Build GRCh38
rs918657685 Chr.18: 24157009-24157009 on Build GRCh38
rs756376903 Chr.18: 24156886-24156886 on Build GRCh38
rs769947094 Chr.18: 24156801-24156801 on Build GRCh38
rs775180815 Chr.18: 24156935-24156935 on Build GRCh38
rs763115014 Chr.18: 24157016-24157017 on Build GRCh38
rs147349985 Chr.18: 24156808-24156808 on Build GRCh38
rs760395784 Chr.18: 24156723-24156735 on Build GRCh38
rs373190949 Chr.18: 24156766-24156766 on Build GRCh38
rs773085151 Chr.18: 24156812-24156812 on Build GRCh38
rs955800232 Chr.18: 24156832-24156832 on Build GRCh38
rs906803437 Chr.18: 24157162-24157162 on Build GRCh38
rs914426763 Chr.18: 24156863-24156863 on Build GRCh38
rs769522646 Chr.18: 24157011-24157011 on Build GRCh38
rs752058549 Chr.18: 24156948-24156948 on Build GRCh38
rs111994740 Chr.18: 24156940-24156940 on Build GRCh38

View Full Product Details