Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111776836-111777365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCND3
Entrez Gene ID
3752
Gene Name
potassium voltage-gated channel subfamily D member 3
Gene Aliases
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22
UniGene
Hs.666367
Gene Chromosome Location
Chr.1: 111770662-111989577 on Build GRCh38
Transcripts
NM_004980.4 NM_172198.2 XM_005270851.4 XM_006710629.3 XM_006710630.3 XM_006710631.3 XM_017001244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471291 c.1727T>C p.I576T Chr.1: 111777065-111777065 on Build GRCh38
3801262 c.1633A>C p.T545P Chr.1: 111777159-111777159 on Build GRCh38
5477460 c.1591C>T p.R531C Chr.1: 111777144-111777144 on Build GRCh38
7336050 c.1619A>T p.N540I Chr.1: 111777116-111777116 on Build GRCh38
4857222 c.1690G>A p.A564T Chr.1: 111777102-111777102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769931414 Chr.1: 111777209-111777209 on Build GRCh38
rs1019786983 Chr.1: 111777055-111777055 on Build GRCh38
rs767169899 Chr.1: 111777001-111777001 on Build GRCh38
rs768952584 Chr.1: 111777231-111777231 on Build GRCh38
rs763028439 Chr.1: 111777077-111777077 on Build GRCh38
rs35027371 Chr.1: 111777146-111777146 on Build GRCh38
rs755768141 Chr.1: 111777017-111777017 on Build GRCh38
rs373808053 Chr.1: 111776991-111776991 on Build GRCh38
rs756541806 Chr.1: 111777323-111777323 on Build GRCh38
rs757809004 Chr.1: 111777164-111777164 on Build GRCh38
rs749559538 Chr.1: 111777228-111777228 on Build GRCh38
rs369415709 Chr.1: 111777000-111777000 on Build GRCh38
rs145890206 Chr.1: 111777219-111777219 on Build GRCh38
rs148478583 Chr.1: 111777067-111777067 on Build GRCh38
rs772508641 Chr.1: 111777288-111777290 on Build GRCh38
rs372635621 Chr.1: 111777007-111777007 on Build GRCh38
rs539783611 Chr.1: 111777232-111777232 on Build GRCh38
rs765916708 Chr.1: 111777268-111777268 on Build GRCh38
rs992442029 Chr.1: 111776863-111776863 on Build GRCh38
rs145710286 Chr.1: 111777173-111777173 on Build GRCh38
rs771385731 Chr.1: 111777040-111777040 on Build GRCh38
rs151164490 Chr.1: 111777143-111777143 on Build GRCh38
rs774340229 Chr.1: 111777238-111777238 on Build GRCh38
rs916805066 Chr.1: 111776865-111776865 on Build GRCh38
rs774064556 Chr.1: 111777260-111777260 on Build GRCh38
rs889011484 Chr.1: 111776927-111776927 on Build GRCh38
rs1050391314 Chr.1: 111776926-111776926 on Build GRCh38
rs925278236 Chr.1: 111777041-111777041 on Build GRCh38
rs923149819 Chr.1: 111776911-111776911 on Build GRCh38
rs879742554 Chr.1: 111776945-111776945 on Build GRCh38
rs762803666 Chr.1: 111776980-111776980 on Build GRCh38
rs547507453 Chr.1: 111777061-111777061 on Build GRCh38
rs867827367 Chr.1: 111777078-111777078 on Build GRCh38
rs1053406521 Chr.1: 111776943-111776943 on Build GRCh38
rs557496343 Chr.1: 111777194-111777194 on Build GRCh38
rs761867267 Chr.1: 111777095-111777095 on Build GRCh38
rs202110939 Chr.1: 111777243-111777243 on Build GRCh38
rs778141653 Chr.1: 111777144-111777144 on Build GRCh38
rs749990612 Chr.1: 111777109-111777109 on Build GRCh38
rs760468985 Chr.1: 111777267-111777267 on Build GRCh38
rs144120746 Chr.1: 111777090-111777090 on Build GRCh38
rs112623380 Chr.1: 111777225-111777225 on Build GRCh38
rs933122273 Chr.1: 111776913-111776913 on Build GRCh38
rs766001729 Chr.1: 111777134-111777134 on Build GRCh38
rs979426177 Chr.1: 111776877-111776877 on Build GRCh38
rs1007503620 Chr.1: 111777046-111777046 on Build GRCh38
rs1035824659 Chr.1: 111776970-111776970 on Build GRCh38
rs751122736 Chr.1: 111776905-111776905 on Build GRCh38
rs969494348 Chr.1: 111777158-111777158 on Build GRCh38
rs1013788086 Chr.1: 111777253-111777253 on Build GRCh38
rs755642943 Chr.1: 111777117-111777117 on Build GRCh38
rs757606694 Chr.1: 111777308-111777308 on Build GRCh38
rs114178167 Chr.1: 111777316-111777316 on Build GRCh38
rs775768536 Chr.1: 111777068-111777068 on Build GRCh38
rs771223175 Chr.1: 111777315-111777315 on Build GRCh38
rs72548732 Chr.1: 111777277-111777277 on Build GRCh38
rs761666801 Chr.1: 111777247-111777247 on Build GRCh38
rs913134158 Chr.1: 111777307-111777307 on Build GRCh38
rs559723424 Chr.1: 111776950-111776950 on Build GRCh38
rs139901716 Chr.1: 111777096-111777096 on Build GRCh38
rs748524463 Chr.1: 111777023-111777023 on Build GRCh38
rs773265114 Chr.1: 111777261-111777261 on Build GRCh38
rs753538536 Chr.1: 111777139-111777139 on Build GRCh38
rs745993925 Chr.1: 111777195-111777195 on Build GRCh38
rs1005880665 Chr.1: 111776944-111776944 on Build GRCh38
rs775978779 Chr.1: 111777291-111777293 on Build GRCh38
rs72548731 Chr.1: 111777340-111777340 on Build GRCh38
rs1011233362 Chr.1: 111776970-111776974 on Build GRCh38
rs200212002 Chr.1: 111777089-111777089 on Build GRCh38
rs1037541026 Chr.1: 111777205-111777205 on Build GRCh38
rs897513806 Chr.1: 111777208-111777208 on Build GRCh38
rs781478104 Chr.1: 111777309-111777309 on Build GRCh38
rs778053688 Chr.1: 111777036-111777036 on Build GRCh38
rs923335129 Chr.1: 111777091-111777091 on Build GRCh38
rs765781656 Chr.1: 111776882-111776882 on Build GRCh38
rs202226567 Chr.1: 111777059-111777059 on Build GRCh38
rs935412719 Chr.1: 111776895-111776895 on Build GRCh38
rs779723268 Chr.1: 111777022-111777022 on Build GRCh38
rs751989149 Chr.1: 111777280-111777280 on Build GRCh38
rs745966594 Chr.1: 111776992-111776993 on Build GRCh38
rs376408039 Chr.1: 111777278-111777278 on Build GRCh38
rs751559149 Chr.1: 111776995-111776995 on Build GRCh38
rs545602928 Chr.1: 111777030-111777030 on Build GRCh38
rs776713733 Chr.1: 111777054-111777054 on Build GRCh38
rs762591204 Chr.1: 111777045-111777045 on Build GRCh38
rs941959013 Chr.1: 111777185-111777185 on Build GRCh38
rs755438937 Chr.1: 111776990-111776990 on Build GRCh38
rs969401702 Chr.1: 111776867-111776867 on Build GRCh38
rs993263038 Chr.1: 111776962-111776962 on Build GRCh38
rs369907159 Chr.1: 111777249-111777249 on Build GRCh38
rs866973745 Chr.1: 111777180-111777180 on Build GRCh38
rs753053248 Chr.1: 111777128-111777128 on Build GRCh38

View Full Product Details