Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67293811-67294313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892289 c.1950G>A p.W650* Chr.16: 67293812-67293812 on Build GRCh38
4129190 c.1877A>C p.D626A Chr.16: 67293885-67293885 on Build GRCh38
225044 c.1867G>A p.E623K Chr.16: 67293895-67293895 on Build GRCh38
3511009 c.1683G>A p.M561I Chr.16: 67294079-67294079 on Build GRCh38
4774333 c.1905C>A p.L635L Chr.16: 67293857-67293857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757430326 Chr.16: 67294076-67294076 on Build GRCh38
rs745605880 Chr.16: 67293878-67293878 on Build GRCh38
rs377568772 Chr.16: 67294036-67294036 on Build GRCh38
rs768569300 Chr.16: 67293987-67293987 on Build GRCh38
rs373975193 Chr.16: 67294025-67294025 on Build GRCh38
rs758373760 Chr.16: 67294213-67294213 on Build GRCh38
rs535438937 Chr.16: 67293916-67293916 on Build GRCh38
rs750873550 Chr.16: 67293891-67293891 on Build GRCh38
rs758471519 Chr.16: 67294057-67294057 on Build GRCh38
rs371196353 Chr.16: 67294189-67294189 on Build GRCh38
rs745730380 Chr.16: 67293948-67293948 on Build GRCh38
rs1008486715 Chr.16: 67294244-67294244 on Build GRCh38
rs767217223 Chr.16: 67293889-67293889 on Build GRCh38
rs779247568 Chr.16: 67293845-67293845 on Build GRCh38
rs778903025 Chr.16: 67294106-67294106 on Build GRCh38
rs754978796 Chr.16: 67293844-67293844 on Build GRCh38
rs1028734319 Chr.16: 67294061-67294061 on Build GRCh38
rs747644390 Chr.16: 67294001-67294001 on Build GRCh38
rs372761904 Chr.16: 67294172-67294172 on Build GRCh38
rs754925791 Chr.16: 67294103-67294103 on Build GRCh38
rs776876507 Chr.16: 67294142-67294142 on Build GRCh38
rs1045594651 Chr.16: 67294167-67294167 on Build GRCh38
rs375131325 Chr.16: 67294223-67294223 on Build GRCh38
rs778761721 Chr.16: 67294205-67294205 on Build GRCh38
rs758181872 Chr.16: 67293860-67293860 on Build GRCh38
rs1047238711 Chr.16: 67294206-67294206 on Build GRCh38
rs748344817 Chr.16: 67293854-67293854 on Build GRCh38
rs779849739 Chr.16: 67293961-67293961 on Build GRCh38
rs201700190 Chr.16: 67293937-67293937 on Build GRCh38
rs557865981 Chr.16: 67294069-67294069 on Build GRCh38
rs373332734 Chr.16: 67294144-67294144 on Build GRCh38
rs979004016 Chr.16: 67294246-67294246 on Build GRCh38
rs199518783 Chr.16: 67293880-67293880 on Build GRCh38
rs769658974 Chr.16: 67294032-67294032 on Build GRCh38
rs554125061 Chr.16: 67293917-67293917 on Build GRCh38
rs758634198 Chr.16: 67293940-67293940 on Build GRCh38
rs577762053 Chr.16: 67294259-67294259 on Build GRCh38
rs373585879 Chr.16: 67293836-67293836 on Build GRCh38
rs770554844 Chr.16: 67294129-67294129 on Build GRCh38
rs1026950784 Chr.16: 67294121-67294121 on Build GRCh38
rs773869253 Chr.16: 67293886-67293886 on Build GRCh38
rs539527603 Chr.16: 67294064-67294064 on Build GRCh38
rs772244334 Chr.16: 67293867-67293867 on Build GRCh38
rs74901862 Chr.16: 67293885-67293885 on Build GRCh38
rs201132591 Chr.16: 67294151-67294151 on Build GRCh38
rs746693583 Chr.16: 67294127-67294127 on Build GRCh38
rs183031746 Chr.16: 67293843-67293843 on Build GRCh38
rs757003395 Chr.16: 67293876-67293876 on Build GRCh38
rs769547268 Chr.16: 67293960-67293960 on Build GRCh38
rs572156950 Chr.16: 67294000-67294000 on Build GRCh38
rs544017674 Chr.16: 67294145-67294145 on Build GRCh38
rs754766688 Chr.16: 67294197-67294197 on Build GRCh38
rs898559457 Chr.16: 67294194-67294194 on Build GRCh38
rs931500891 Chr.16: 67294204-67294204 on Build GRCh38
rs748114516 Chr.16: 67294212-67294212 on Build GRCh38
rs757433424 Chr.16: 67294175-67294175 on Build GRCh38
rs763071577 Chr.16: 67294161-67294161 on Build GRCh38
rs1019843710 Chr.16: 67294255-67294255 on Build GRCh38
rs754281134 Chr.16: 67293910-67293910 on Build GRCh38
rs199914291 Chr.16: 67294109-67294109 on Build GRCh38
rs779890887 Chr.16: 67293877-67293878 on Build GRCh38
rs541401542 Chr.16: 67294224-67294224 on Build GRCh38
rs753638911 Chr.16: 67294190-67294190 on Build GRCh38
rs762572868 Chr.16: 67293882-67293882 on Build GRCh38
rs769946132 Chr.16: 67293885-67293888 on Build GRCh38
rs780690255 Chr.16: 67293839-67293839 on Build GRCh38
rs866254895 Chr.16: 67293963-67293963 on Build GRCh38
rs527316881 Chr.16: 67294271-67294271 on Build GRCh38
rs775309307 Chr.16: 67294073-67294073 on Build GRCh38
rs777429162 Chr.16: 67294220-67294220 on Build GRCh38
rs759968343 Chr.16: 67294029-67294029 on Build GRCh38
rs773746023 Chr.16: 67293988-67293988 on Build GRCh38
rs771683395 Chr.16: 67294118-67294118 on Build GRCh38
rs779715634 Chr.16: 67294098-67294098 on Build GRCh38
rs766679578 Chr.16: 67294024-67294024 on Build GRCh38
rs1018639461 Chr.16: 67294027-67294027 on Build GRCh38
rs777850527 Chr.16: 67293935-67293936 on Build GRCh38
rs749324566 Chr.16: 67294166-67294167 on Build GRCh38
rs200302400 Chr.16: 67293881-67293881 on Build GRCh38
rs761112874 Chr.16: 67293902-67293902 on Build GRCh38
rs771914695 Chr.16: 67294013-67294013 on Build GRCh38
rs936441491 Chr.16: 67293968-67293968 on Build GRCh38
rs1045118742 Chr.16: 67293853-67293853 on Build GRCh38
rs780973499 Chr.16: 67293877-67293877 on Build GRCh38
rs752833909 Chr.16: 67294056-67294056 on Build GRCh38
rs936305690 Chr.16: 67294270-67294270 on Build GRCh38
rs772964262 Chr.16: 67294020-67294020 on Build GRCh38
rs200635073 Chr.16: 67293943-67293943 on Build GRCh38
rs748374625 Chr.16: 67293885-67293889 on Build GRCh38
rs749215254 Chr.16: 67293962-67293962 on Build GRCh38
rs560071752 Chr.16: 67294248-67294248 on Build GRCh38
rs772804024 Chr.16: 67294120-67294120 on Build GRCh38
rs576197532 Chr.16: 67294100-67294100 on Build GRCh38
rs779125067 Chr.16: 67294037-67294037 on Build GRCh38

View Full Product Details