Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64271602-64271855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAD
Entrez Gene ID
572
Gene Name
BCL2 associated agonist of cell death
Gene Aliases
BBC2, BCL2L8
UniGene
Hs.370254
Gene Chromosome Location
Chr.11: 64269828-64284704 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018016.1

Gene Symbol
GPR137
Entrez Gene ID
56834
Gene Name
G protein-coupled receptor 137
Gene Aliases
C11orf4, GPR137A, TM7SF1L1
UniGene
Hs.523763
Gene Chromosome Location
Chr.11: 64270337-64289500 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39869 c.371C>T p.S124F Chr.11: 64271620-64271620 on Build GRCh38
1184361 c.301C>T p.P101S Chr.11: 64271690-64271690 on Build GRCh38
930061 c.298G>A p.A100T Chr.11: 64271693-64271693 on Build GRCh38
6312416 c.306C>A p.P102P Chr.11: 64271685-64271685 on Build GRCh38
930062 c.243G>A p.E81E Chr.11: 64271748-64271748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756609479 Chr.11: 64271794-64271794 on Build GRCh38
rs570577158 Chr.11: 64271719-64271719 on Build GRCh38
rs767611787 Chr.11: 64271785-64271785 on Build GRCh38
rs927566353 Chr.11: 64271634-64271634 on Build GRCh38
rs199786293 Chr.11: 64271667-64271667 on Build GRCh38
rs554645051 Chr.11: 64271758-64271758 on Build GRCh38
rs375432562 Chr.11: 64271736-64271736 on Build GRCh38
rs763267746 Chr.11: 64271660-64271660 on Build GRCh38
rs546500553 Chr.11: 64271738-64271738 on Build GRCh38
rs534875657 Chr.11: 64271625-64271625 on Build GRCh38
rs368351638 Chr.11: 64271775-64271775 on Build GRCh38
rs756625685 Chr.11: 64271685-64271685 on Build GRCh38
rs769442984 Chr.11: 64271830-64271830 on Build GRCh38
rs375657682 Chr.11: 64271682-64271682 on Build GRCh38
rs1023100466 Chr.11: 64271777-64271777 on Build GRCh38
rs780382977 Chr.11: 64271694-64271694 on Build GRCh38
rs963571381 Chr.11: 64271784-64271784 on Build GRCh38
rs750524708 Chr.11: 64271790-64271790 on Build GRCh38
rs777345859 Chr.11: 64271733-64271733 on Build GRCh38
rs995520756 Chr.11: 64271806-64271806 on Build GRCh38
rs374273002 Chr.11: 64271773-64271773 on Build GRCh38
rs943224621 Chr.11: 64271678-64271678 on Build GRCh38
rs776579950 Chr.11: 64271621-64271621 on Build GRCh38
rs2286615 Chr.11: 64271703-64271703 on Build GRCh38
rs959001896 Chr.11: 64271638-64271638 on Build GRCh38
rs574741077 Chr.11: 64271816-64271816 on Build GRCh38
rs902202934 Chr.11: 64271714-64271714 on Build GRCh38
rs2286616 Chr.11: 64271664-64271664 on Build GRCh38
rs762780261 Chr.11: 64271750-64271750 on Build GRCh38
rs368660221 Chr.11: 64271747-64271747 on Build GRCh38
rs919269601 Chr.11: 64271629-64271629 on Build GRCh38
rs745310593 Chr.11: 64271829-64271829 on Build GRCh38
rs746653543 Chr.11: 64271820-64271820 on Build GRCh38
rs3729933 Chr.11: 64271672-64271672 on Build GRCh38
rs754351648 Chr.11: 64271802-64271802 on Build GRCh38
rs752601428 Chr.11: 64271643-64271643 on Build GRCh38
rs759434976 Chr.11: 64271639-64271639 on Build GRCh38
rs192629069 Chr.11: 64271655-64271655 on Build GRCh38
rs780468801 Chr.11: 64271795-64271795 on Build GRCh38
rs1038889519 Chr.11: 64271681-64271681 on Build GRCh38
rs780752598 Chr.11: 64271825-64271825 on Build GRCh38
rs755283304 Chr.11: 64271809-64271809 on Build GRCh38
rs920264781 Chr.11: 64271671-64271671 on Build GRCh38
rs776057256 Chr.11: 64271742-64271742 on Build GRCh38
rs987988841 Chr.11: 64271646-64271646 on Build GRCh38
rs145689917 Chr.11: 64271745-64271745 on Build GRCh38
rs769010021 Chr.11: 64271726-64271726 on Build GRCh38
rs748893784 Chr.11: 64271728-64271729 on Build GRCh38
rs370969520 Chr.11: 64271748-64271748 on Build GRCh38
rs756919548 Chr.11: 64271823-64271823 on Build GRCh38
rs781162957 Chr.11: 64271679-64271679 on Build GRCh38
rs765104579 Chr.11: 64271641-64271641 on Build GRCh38
rs764003005 Chr.11: 64271751-64271751 on Build GRCh38
rs757313178 Chr.11: 64271675-64271675 on Build GRCh38

View Full Product Details